Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3500927-3501182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUAP1
Entrez Gene ID
23059
Gene Name
clusterin associated protein 1
Gene Aliases
CFAP22, FAP22, IFT38
UniGene
Hs.155995
Gene Chromosome Location
Chr.16: 3495562-3539048 on Build GRCh38
Transcripts
NM_015041.2 XM_006720867.3 XM_017023068.1 XM_017023069.1 XM_017023070.1 XM_017023071.1 XM_017023072.1 XM_017023073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389262 c.22A>G p.N8D Chr.16: 3501089-3501089 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs115697151 Chr.16: 3501143-3501143 on Build GRCh38
rs1046768016 Chr.16: 3501003-3501003 on Build GRCh38
rs756063098 Chr.16: 3501119-3501119 on Build GRCh38
rs752625438 Chr.16: 3501116-3501116 on Build GRCh38
rs749076413 Chr.16: 3501141-3501141 on Build GRCh38
rs762646838 Chr.16: 3501102-3501102 on Build GRCh38
rs146525471 Chr.16: 3501011-3501011 on Build GRCh38
rs774456533 Chr.16: 3501068-3501068 on Build GRCh38
rs575259168 Chr.16: 3501083-3501083 on Build GRCh38
rs371966920 Chr.16: 3501051-3501051 on Build GRCh38
rs111497094 Chr.16: 3500978-3500978 on Build GRCh38
rs79384355 Chr.16: 3500998-3500998 on Build GRCh38
rs372807107 Chr.16: 3501029-3501029 on Build GRCh38
rs777495289 Chr.16: 3501057-3501057 on Build GRCh38
rs745977326 Chr.16: 3501076-3501076 on Build GRCh38
rs867829100 Chr.16: 3501151-3501151 on Build GRCh38
rs78552393 Chr.16: 3501067-3501067 on Build GRCh38
rs562071429 Chr.16: 3500999-3500999 on Build GRCh38
rs201752743 Chr.16: 3501107-3501107 on Build GRCh38
rs535132010 Chr.16: 3501055-3501055 on Build GRCh38
rs1007660932 Chr.16: 3501030-3501030 on Build GRCh38
rs761143695 Chr.16: 3501028-3501028 on Build GRCh38
rs375226001 Chr.16: 3501053-3501053 on Build GRCh38
rs964248394 Chr.16: 3501153-3501153 on Build GRCh38
rs751121956 Chr.16: 3501043-3501043 on Build GRCh38
rs1011257599 Chr.16: 3501121-3501121 on Build GRCh38
rs375774922 Chr.16: 3501034-3501034 on Build GRCh38
rs750097689 Chr.16: 3501032-3501032 on Build GRCh38
rs202030605 Chr.16: 3501046-3501046 on Build GRCh38
rs1054690926 Chr.16: 3500952-3500952 on Build GRCh38
rs1051935372 Chr.16: 3501008-3501008 on Build GRCh38
rs776923266 Chr.16: 3501091-3501091 on Build GRCh38
rs374465696 Chr.16: 3500997-3500997 on Build GRCh38
rs369602114 Chr.16: 3501096-3501096 on Build GRCh38
rs765901773 Chr.16: 3501038-3501038 on Build GRCh38
rs534333509 Chr.16: 3501084-3501084 on Build GRCh38
rs796202431 Chr.16: 3500955-3500955 on Build GRCh38
rs373061370 Chr.16: 3501103-3501103 on Build GRCh38
rs777599977 Chr.16: 3501122-3501122 on Build GRCh38
rs531119586 Chr.16: 3501000-3501000 on Build GRCh38
rs772659372 Chr.16: 3501081-3501081 on Build GRCh38
rs754494832 Chr.16: 3501109-3501109 on Build GRCh38
rs937772031 Chr.16: 3500950-3500950 on Build GRCh38
rs919462672 Chr.16: 3501105-3501105 on Build GRCh38
rs2301762 Chr.16: 3500977-3500977 on Build GRCh38
rs368038749 Chr.16: 3501149-3501149 on Build GRCh38
rs755543900 Chr.16: 3501068-3501068 on Build GRCh38
rs369728865 Chr.16: 3501112-3501112 on Build GRCh38
rs749512422 Chr.16: 3501058-3501058 on Build GRCh38
rs752461135 Chr.16: 3501048-3501049 on Build GRCh38
rs1017579923 Chr.16: 3501033-3501033 on Build GRCh38
rs775959509 Chr.16: 3501026-3501026 on Build GRCh38
rs897983718 Chr.16: 3500982-3500982 on Build GRCh38
rs771106081 Chr.16: 3501061-3501061 on Build GRCh38
rs959093385 Chr.16: 3501082-3501082 on Build GRCh38
rs1037952788 Chr.16: 3501104-3501104 on Build GRCh38
rs758131441 Chr.16: 3501087-3501087 on Build GRCh38

View Full Product Details