Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133586662-133586887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDV3
Entrez Gene ID
55573
Gene Name
CDV3 homolog
Gene Aliases
H41
UniGene
Hs.518265
Gene Chromosome Location
Chr.3: 133573563-133590274 on Build GRCh38
Transcripts
NM_001134422.1 NM_001134423.2 NM_001282762.1 NM_001282763.1 NM_001282764.1 NM_001282765.1 NM_017548.4 XM_005247588.4 XM_005247589.4 XM_005247590.4 XM_005247591.4 XM_005247592.4 XM_005247594.4 XM_011512961.2 XM_011512962.2 XM_017006808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033467 c.587C>A p.P196Q Chr.3: 133586683-133586683 on Build GRCh38
5863550 c.572G>A p.S191N Chr.3: 133586668-133586668 on Build GRCh38
5863551 c.572G>A p.S191N Chr.3: 133586668-133586668 on Build GRCh38
6033468 c.587C>A p.P196Q Chr.3: 133586683-133586683 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901432179 Chr.3: 133586729-133586729 on Build GRCh38
rs112389024 Chr.3: 133586767-133586767 on Build GRCh38
rs779223581 Chr.3: 133586765-133586765 on Build GRCh38
rs1016756792 Chr.3: 133586824-133586824 on Build GRCh38
rs913022213 Chr.3: 133586778-133586778 on Build GRCh38
rs760687869 Chr.3: 133586748-133586748 on Build GRCh38
rs927868127 Chr.3: 133586852-133586852 on Build GRCh38
rs374297540 Chr.3: 133586718-133586718 on Build GRCh38
rs972637205 Chr.3: 133586848-133586848 on Build GRCh38
rs774338355 Chr.3: 133586713-133586713 on Build GRCh38
rs547179516 Chr.3: 133586732-133586732 on Build GRCh38
rs529564727 Chr.3: 133586822-133586822 on Build GRCh38
rs201471631 Chr.3: 133586765-133586765 on Build GRCh38
rs1053409048 Chr.3: 133586704-133586704 on Build GRCh38
rs1006497154 Chr.3: 133586758-133586758 on Build GRCh38
rs189060779 Chr.3: 133586835-133586835 on Build GRCh38
rs754005069 Chr.3: 133586752-133586752 on Build GRCh38
rs968579117 Chr.3: 133586849-133586849 on Build GRCh38
rs773081898 Chr.3: 133586737-133586738 on Build GRCh38
rs560788665 Chr.3: 133586818-133586818 on Build GRCh38
rs988581115 Chr.3: 133586769-133586769 on Build GRCh38
rs541142196 Chr.3: 133586739-133586742 on Build GRCh38
rs767595006 Chr.3: 133586730-133586730 on Build GRCh38
rs978615917 Chr.3: 133586858-133586858 on Build GRCh38
rs150055916 Chr.3: 133586862-133586862 on Build GRCh38
rs961238083 Chr.3: 133586832-133586832 on Build GRCh38
rs999062789 Chr.3: 133586697-133586697 on Build GRCh38
rs1005666398 Chr.3: 133586779-133586779 on Build GRCh38
rs1028759635 Chr.3: 133586760-133586760 on Build GRCh38
rs764295624 Chr.3: 133586751-133586751 on Build GRCh38

View Full Product Details