Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121090208-121090469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
UniGene
Hs.653263
Gene Chromosome Location
Chr.9: 121074778-121177610 on Build GRCh38
Transcripts
NM_007018.4 XM_005251679.3 XM_006716940.2 XM_006716944.3 XM_011518166.1 XM_011518167.1 XM_011518168.1 XM_011518169.2 XM_011518170.1 XM_011518172.2 XM_017014223.1 XM_017014224.1 XM_017014225.1 XM_017014227.1 XM_017014228.1 XM_017014229.1 XM_017014230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314413 c.339G>A p.K113K Chr.9: 121090396-121090396 on Build GRCh38
310046 c.297G>T p.L99F Chr.9: 121090354-121090354 on Build GRCh38
1624493 c.291G>T p.L97F Chr.9: 121090348-121090348 on Build GRCh38
5100217 c.309G>T p.L103L Chr.9: 121090366-121090366 on Build GRCh38
7176866 c.307C>G p.L103V Chr.9: 121090364-121090364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772581542 Chr.9: 121090411-121090411 on Build GRCh38
rs775053654 Chr.9: 121090391-121090391 on Build GRCh38
rs767836751 Chr.9: 121090290-121090290 on Build GRCh38
rs779390776 Chr.9: 121090293-121090293 on Build GRCh38
rs764107634 Chr.9: 121090300-121090300 on Build GRCh38
rs752869551 Chr.9: 121090295-121090295 on Build GRCh38
rs775735949 Chr.9: 121090420-121090420 on Build GRCh38
rs887980364 Chr.9: 121090252-121090252 on Build GRCh38
rs1007507571 Chr.9: 121090419-121090419 on Build GRCh38
rs760842261 Chr.9: 121090436-121090436 on Build GRCh38
rs758254553 Chr.9: 121090353-121090353 on Build GRCh38
rs753769506 Chr.9: 121090306-121090306 on Build GRCh38
rs746654354 Chr.9: 121090366-121090366 on Build GRCh38
rs779613516 Chr.9: 121090357-121090357 on Build GRCh38
rs777887928 Chr.9: 121090345-121090345 on Build GRCh38
rs773709818 Chr.9: 121090240-121090240 on Build GRCh38
rs766805179 Chr.9: 121090270-121090270 on Build GRCh38
rs1009983073 Chr.9: 121090340-121090340 on Build GRCh38
rs140149582 Chr.9: 121090388-121090388 on Build GRCh38
rs559706427 Chr.9: 121090410-121090410 on Build GRCh38
rs564006480 Chr.9: 121090324-121090324 on Build GRCh38
rs760023372 Chr.9: 121090288-121090288 on Build GRCh38
rs768162564 Chr.9: 121090384-121090384 on Build GRCh38
rs750208625 Chr.9: 121090331-121090331 on Build GRCh38
rs376461563 Chr.9: 121090278-121090278 on Build GRCh38
rs117390062 Chr.9: 121090387-121090387 on Build GRCh38
rs144423052 Chr.9: 121090333-121090333 on Build GRCh38
rs201460318 Chr.9: 121090399-121090399 on Build GRCh38
rs897580510 Chr.9: 121090351-121090351 on Build GRCh38
rs370010588 Chr.9: 121090322-121090322 on Build GRCh38
rs760753373 Chr.9: 121090298-121090298 on Build GRCh38
rs763392494 Chr.9: 121090265-121090265 on Build GRCh38

View Full Product Details