Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10126148-10126409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
UniGene
Hs.649307
Gene Chromosome Location
Chr.3: 10115649-10127190 on Build GRCh38
Transcripts
NM_018462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3845694 c.297C>T p.F99F Chr.3: 10126368-10126368 on Build GRCh38
222776 c.202-1G>A p.? Chr.3: 10126268-10126268 on Build GRCh38
3364914 c.208A>C p.K70Q Chr.3: 10126275-10126275 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564692778 Chr.3: 10126222-10126222 on Build GRCh38
rs1023969671 Chr.3: 10126203-10126203 on Build GRCh38
rs181193811 Chr.3: 10126189-10126189 on Build GRCh38
rs767554967 Chr.3: 10126241-10126241 on Build GRCh38
rs781407599 Chr.3: 10126346-10126346 on Build GRCh38
rs750585206 Chr.3: 10126305-10126305 on Build GRCh38
rs533510454 Chr.3: 10126384-10126384 on Build GRCh38
rs868665556 Chr.3: 10126298-10126298 on Build GRCh38
rs754083550 Chr.3: 10126289-10126289 on Build GRCh38
rs982175006 Chr.3: 10126193-10126193 on Build GRCh38
rs184954016 Chr.3: 10126191-10126191 on Build GRCh38
rs765238543 Chr.3: 10126304-10126304 on Build GRCh38
rs756849074 Chr.3: 10126223-10126236 on Build GRCh38
rs370276554 Chr.3: 10126248-10126248 on Build GRCh38
rs564589910 Chr.3: 10126202-10126203 on Build GRCh38
rs908201804 Chr.3: 10126344-10126344 on Build GRCh38
rs775250491 Chr.3: 10126243-10126243 on Build GRCh38
rs374564323 Chr.3: 10126174-10126174 on Build GRCh38
rs759394013 Chr.3: 10126225-10126225 on Build GRCh38
rs927949092 Chr.3: 10126215-10126215 on Build GRCh38
rs879786082 Chr.3: 10126205-10126205 on Build GRCh38
rs758821709 Chr.3: 10126322-10126322 on Build GRCh38
rs764885813 Chr.3: 10126233-10126234 on Build GRCh38
rs554914757 Chr.3: 10126216-10126216 on Build GRCh38
rs777514824 Chr.3: 10126377-10126377 on Build GRCh38
rs866753021 Chr.3: 10126267-10126267 on Build GRCh38
rs747142969 Chr.3: 10126220-10126220 on Build GRCh38
rs761824971 Chr.3: 10126300-10126300 on Build GRCh38
rs777647053 Chr.3: 10126217-10126217 on Build GRCh38
rs972900962 Chr.3: 10126378-10126378 on Build GRCh38
rs910073386 Chr.3: 10126324-10126324 on Build GRCh38
rs551136546 Chr.3: 10126188-10126188 on Build GRCh38
rs755421081 Chr.3: 10126368-10126368 on Build GRCh38
rs866567720 Chr.3: 10126314-10126314 on Build GRCh38
rs572806196 Chr.3: 10126249-10126249 on Build GRCh38
rs918301463 Chr.3: 10126380-10126380 on Build GRCh38
rs34046803 Chr.3: 10126315-10126316 on Build GRCh38
rs780664282 Chr.3: 10126363-10126363 on Build GRCh38
rs749917328 Chr.3: 10126254-10126254 on Build GRCh38
rs745993527 Chr.3: 10126364-10126364 on Build GRCh38

View Full Product Details