Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13319233-13319498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3L
Entrez Gene ID
92552
Gene Name
ataxin 3 like
Gene Aliases
MJDL
UniGene
Hs.382641
Gene Chromosome Location
Chr.X: 13318647-13320399 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

Gene Symbol
GS1-600G8.3
Entrez Gene ID
100093698
Gene Name
unknown transcript
Gene Aliases
-
UniGene
Hs.643612
Gene Chromosome Location
Chr.X: 13310652-13319933 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

SNP Detail

SNP ID Chromosome Location
rs768318062 Chr.X: 13319295-13319295 on Build GRCh38
rs767792784 Chr.X: 13319356-13319356 on Build GRCh38
rs772727645 Chr.X: 13319463-13319463 on Build GRCh38
rs746865828 Chr.X: 13319282-13319282 on Build GRCh38
rs968120770 Chr.X: 13319299-13319299 on Build GRCh38
rs779809245 Chr.X: 13319452-13319452 on Build GRCh38
rs199943946 Chr.X: 13319441-13319441 on Build GRCh38
rs201093759 Chr.X: 13319329-13319329 on Build GRCh38
rs759596951 Chr.X: 13319350-13319350 on Build GRCh38
rs199954866 Chr.X: 13319444-13319444 on Build GRCh38
rs780851738 Chr.X: 13319459-13319459 on Build GRCh38
rs779851334 Chr.X: 13319273-13319273 on Build GRCh38
rs992449399 Chr.X: 13319399-13319399 on Build GRCh38
rs772585490 Chr.X: 13319292-13319293 on Build GRCh38
rs762425230 Chr.X: 13319414-13319414 on Build GRCh38
rs367551011 Chr.X: 13319385-13319385 on Build GRCh38
rs773688226 Chr.X: 13319461-13319463 on Build GRCh38
rs774425178 Chr.X: 13319348-13319348 on Build GRCh38
rs747877809 Chr.X: 13319466-13319466 on Build GRCh38
rs769649021 Chr.X: 13319344-13319344 on Build GRCh38
rs765827656 Chr.X: 13319420-13319420 on Build GRCh38
rs751190262 Chr.X: 13319458-13319458 on Build GRCh38
rs754165902 Chr.X: 13319386-13319386 on Build GRCh38
rs373746855 Chr.X: 13319370-13319370 on Build GRCh38
rs762167713 Chr.X: 13319257-13319257 on Build GRCh38
rs776434126 Chr.X: 13319298-13319298 on Build GRCh38
rs201089107 Chr.X: 13319306-13319306 on Build GRCh38
rs769343025 Chr.X: 13319382-13319382 on Build GRCh38
rs745317447 Chr.X: 13319266-13319266 on Build GRCh38
rs868368488 Chr.X: 13319436-13319436 on Build GRCh38
rs376294771 Chr.X: 13319474-13319474 on Build GRCh38
rs367726512 Chr.X: 13319269-13319269 on Build GRCh38
rs747767169 Chr.X: 13319378-13319378 on Build GRCh38

View Full Product Details