Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93203611-93203872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GON7
Entrez Gene ID
84520
Gene Name
GON7, KEOPS complex subunit homolog
Gene Aliases
C14orf142, PNAS-127
UniGene
Hs.20142
Gene Chromosome Location
Chr.14: 93202892-93207114 on Build GRCh38
Transcripts
NM_032490.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666840 c.283C>T p.R95W Chr.14: 93203708-93203708 on Build GRCh38
6880373 c.287C>T p.P96L Chr.14: 93203704-93203704 on Build GRCh38
6614285 c.209-2A>G p.? Chr.14: 93203784-93203784 on Build GRCh38
958928 c.226G>A p.A76T Chr.14: 93203765-93203765 on Build GRCh38
958927 c.264C>T p.F88F Chr.14: 93203727-93203727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535902711 Chr.14: 93203645-93203645 on Build GRCh38
rs1041465799 Chr.14: 93203674-93203674 on Build GRCh38
rs773640502 Chr.14: 93203686-93203686 on Build GRCh38
rs748197151 Chr.14: 93203782-93203782 on Build GRCh38
rs748761288 Chr.14: 93203656-93203656 on Build GRCh38
rs769420766 Chr.14: 93203643-93203643 on Build GRCh38
rs534896711 Chr.14: 93203708-93203708 on Build GRCh38
rs778492301 Chr.14: 93203795-93203795 on Build GRCh38
rs750224435 Chr.14: 93203748-93203748 on Build GRCh38
rs1026317895 Chr.14: 93203819-93203819 on Build GRCh38
rs748547172 Chr.14: 93203799-93203799 on Build GRCh38
rs766704950 Chr.14: 93203695-93203695 on Build GRCh38
rs114819010 Chr.14: 93203694-93203694 on Build GRCh38
rs1049963187 Chr.14: 93203788-93203788 on Build GRCh38
rs757229683 Chr.14: 93203733-93203733 on Build GRCh38
rs993560347 Chr.14: 93203818-93203818 on Build GRCh38
rs772261056 Chr.14: 93203682-93203682 on Build GRCh38
rs369111205 Chr.14: 93203815-93203815 on Build GRCh38
rs779654694 Chr.14: 93203765-93203765 on Build GRCh38
rs772620723 Chr.14: 93203803-93203803 on Build GRCh38
rs908460781 Chr.14: 93203644-93203644 on Build GRCh38
rs890852816 Chr.14: 93203796-93203796 on Build GRCh38
rs781046307 Chr.14: 93203734-93203734 on Build GRCh38
rs776952455 Chr.14: 93203829-93203829 on Build GRCh38
rs755939301 Chr.14: 93203763-93203763 on Build GRCh38
rs1019349033 Chr.14: 93203797-93203797 on Build GRCh38
rs943711841 Chr.14: 93203805-93203805 on Build GRCh38
rs754152964 Chr.14: 93203697-93203697 on Build GRCh38
rs771760136 Chr.14: 93203747-93203747 on Build GRCh38
rs760240597 Chr.14: 93203810-93203810 on Build GRCh38
rs774675117 Chr.14: 93203676-93203676 on Build GRCh38
rs773660146 Chr.14: 93203804-93203804 on Build GRCh38
rs1054743933 Chr.14: 93203743-93203743 on Build GRCh38
rs555105190 Chr.14: 93203782-93203782 on Build GRCh38
rs772995650 Chr.14: 93203827-93203830 on Build GRCh38
rs1007581194 Chr.14: 93203784-93203784 on Build GRCh38
rs779744808 Chr.14: 93203648-93203648 on Build GRCh38
rs892760637 Chr.14: 93203821-93203821 on Build GRCh38
rs951225233 Chr.14: 93203837-93203837 on Build GRCh38
rs771444436 Chr.14: 93203816-93203816 on Build GRCh38
rs182594724 Chr.14: 93203660-93203660 on Build GRCh38
rs765437405 Chr.14: 93203711-93203711 on Build GRCh38
rs973939467 Chr.14: 93203672-93203672 on Build GRCh38
rs372149848 Chr.14: 93203681-93203681 on Build GRCh38
rs1009766828 Chr.14: 93203836-93203836 on Build GRCh38
rs751384720 Chr.14: 93203727-93203727 on Build GRCh38
rs1000334105 Chr.14: 93203707-93203707 on Build GRCh38

View Full Product Details