Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203683153-203683426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
UniGene
Hs.343522
Gene Chromosome Location
Chr.1: 203626787-203744081 on Build GRCh38
Transcripts
NM_001001396.2 NM_001684.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6124135 c.167G>A p.S56N Chr.1: 203683372-203683372 on Build GRCh38
6060991 c.162C>G p.L54L Chr.1: 203683367-203683367 on Build GRCh38
7180093 c.96C>T p.L32L Chr.1: 203683301-203683301 on Build GRCh38
6060993 c.146G>T p.G49V Chr.1: 203683351-203683351 on Build GRCh38
1601541 c.153A>G p.V51V Chr.1: 203683358-203683358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936312041 Chr.1: 203683283-203683283 on Build GRCh38
rs777078203 Chr.1: 203683233-203683233 on Build GRCh38
rs375654382 Chr.1: 203683307-203683307 on Build GRCh38
rs371098430 Chr.1: 203683354-203683354 on Build GRCh38
rs202241442 Chr.1: 203683275-203683275 on Build GRCh38
rs373427232 Chr.1: 203683377-203683377 on Build GRCh38
rs923644490 Chr.1: 203683279-203683279 on Build GRCh38
rs758076020 Chr.1: 203683185-203683185 on Build GRCh38
rs758519874 Chr.1: 203683269-203683269 on Build GRCh38
rs377159132 Chr.1: 203683181-203683181 on Build GRCh38
rs778798145 Chr.1: 203683265-203683265 on Build GRCh38
rs763740976 Chr.1: 203683194-203683194 on Build GRCh38
rs771059469 Chr.1: 203683258-203683258 on Build GRCh38
rs377401279 Chr.1: 203683389-203683389 on Build GRCh38
rs142598582 Chr.1: 203683250-203683250 on Build GRCh38
rs751719143 Chr.1: 203683399-203683399 on Build GRCh38
rs760514100 Chr.1: 203683346-203683346 on Build GRCh38
rs776509703 Chr.1: 203683262-203683262 on Build GRCh38
rs779554688 Chr.1: 203683201-203683201 on Build GRCh38
rs776505845 Chr.1: 203683342-203683343 on Build GRCh38
rs1030977850 Chr.1: 203683266-203683266 on Build GRCh38
rs750242599 Chr.1: 203683345-203683345 on Build GRCh38
rs61731222 Chr.1: 203683341-203683341 on Build GRCh38
rs772349464 Chr.1: 203683218-203683218 on Build GRCh38
rs373387195 Chr.1: 203683200-203683200 on Build GRCh38
rs758426870 Chr.1: 203683365-203683365 on Build GRCh38
rs369212336 Chr.1: 203683251-203683251 on Build GRCh38
rs369576144 Chr.1: 203683197-203683197 on Build GRCh38
rs752876520 Chr.1: 203683267-203683267 on Build GRCh38
rs762562774 Chr.1: 203683312-203683312 on Build GRCh38
rs1419114 Chr.1: 203683316-203683316 on Build GRCh38
rs375107873 Chr.1: 203683225-203683225 on Build GRCh38
rs761589301 Chr.1: 203683325-203683325 on Build GRCh38
rs771250580 Chr.1: 203683292-203683292 on Build GRCh38
rs779076905 Chr.1: 203683211-203683211 on Build GRCh38
rs751460668 Chr.1: 203683195-203683195 on Build GRCh38
rs751366917 Chr.1: 203683257-203683257 on Build GRCh38
rs146234576 Chr.1: 203683224-203683224 on Build GRCh38
rs760120643 Chr.1: 203683243-203683243 on Build GRCh38
rs3766753 Chr.1: 203683177-203683177 on Build GRCh38
rs748248643 Chr.1: 203683217-203683217 on Build GRCh38
rs747258267 Chr.1: 203683280-203683280 on Build GRCh38
rs781541528 Chr.1: 203683293-203683293 on Build GRCh38
rs140289832 Chr.1: 203683210-203683210 on Build GRCh38
rs774860823 Chr.1: 203683311-203683311 on Build GRCh38
rs527900427 Chr.1: 203683246-203683246 on Build GRCh38
rs963788724 Chr.1: 203683335-203683335 on Build GRCh38
rs768293756 Chr.1: 203683206-203683211 on Build GRCh38
rs764586158 Chr.1: 203683244-203683244 on Build GRCh38
rs774096766 Chr.1: 203683320-203683320 on Build GRCh38
rs986432272 Chr.1: 203683315-203683315 on Build GRCh38
rs746304623 Chr.1: 203683302-203683302 on Build GRCh38
rs935270974 Chr.1: 203683356-203683356 on Build GRCh38

View Full Product Details