Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102602070-102602319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5
Entrez Gene ID
64860
Gene Name
armadillo repeat containing, X-linked 5
Gene Aliases
GASP5
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599168-102604159 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

SNP Detail

SNP ID Chromosome Location
rs765525124 Chr.X: 102602128-102602128 on Build GRCh38
rs777165273 Chr.X: 102602252-102602252 on Build GRCh38
rs147081834 Chr.X: 102602167-102602167 on Build GRCh38
rs771504425 Chr.X: 102602106-102602106 on Build GRCh38
rs780086263 Chr.X: 102602125-102602126 on Build GRCh38
rs772283056 Chr.X: 102602201-102602201 on Build GRCh38
rs752178410 Chr.X: 102602160-102602160 on Build GRCh38
rs778633013 Chr.X: 102602250-102602250 on Build GRCh38
rs371888083 Chr.X: 102602100-102602100 on Build GRCh38
rs376521671 Chr.X: 102602151-102602151 on Build GRCh38
rs756396377 Chr.X: 102602249-102602249 on Build GRCh38
rs747687830 Chr.X: 102602105-102602105 on Build GRCh38
rs774185953 Chr.X: 102602127-102602127 on Build GRCh38
rs1017659655 Chr.X: 102602247-102602247 on Build GRCh38
rs17854978 Chr.X: 102602297-102602297 on Build GRCh38
rs757964410 Chr.X: 102602290-102602290 on Build GRCh38
rs1056732662 Chr.X: 102602154-102602154 on Build GRCh38
rs746940067 Chr.X: 102602108-102602108 on Build GRCh38
rs767819081 Chr.X: 102602191-102602191 on Build GRCh38
rs1043865370 Chr.X: 102602113-102602113 on Build GRCh38
rs775783944 Chr.X: 102602146-102602146 on Build GRCh38
rs1012372121 Chr.X: 102602186-102602186 on Build GRCh38
rs747742515 Chr.X: 102602275-102602275 on Build GRCh38
rs764256029 Chr.X: 102602159-102602159 on Build GRCh38
rs770701406 Chr.X: 102602109-102602109 on Build GRCh38

View Full Product Details