Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47157728-47158001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
UniGene
Hs.368296
Gene Chromosome Location
Chr.11: 46936524-47164381 on Build GRCh38
Transcripts
NM_001003676.2 NM_001003677.2 NM_001003678.2 NM_001278222.1 NM_024113.4 NR_103471.1 NR_103472.1 XM_006718315.1 XM_011520364.2 XM_011520365.2 XM_017018276.1 XM_017018277.1 XM_017018278.1 XM_017018279.1 XM_017018280.1 XM_017018281.1 XM_017018282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666641 c.742A>C p.I248L Chr.11: 47157907-47157907 on Build GRCh38
7175453 c.728C>T p.S243L Chr.11: 47157893-47157893 on Build GRCh38
3980344 c.737G>A p.R246H Chr.11: 47157902-47157902 on Build GRCh38
7175451 c.693G>A p.Q231Q Chr.11: 47157858-47157858 on Build GRCh38
4165796 c.730A>T p.K244* Chr.11: 47157895-47157895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915459704 Chr.11: 47157839-47157839 on Build GRCh38
rs751265761 Chr.11: 47157898-47157898 on Build GRCh38
rs779569704 Chr.11: 47157885-47157885 on Build GRCh38
rs779468787 Chr.11: 47157785-47157785 on Build GRCh38
rs753597465 Chr.11: 47157941-47157941 on Build GRCh38
rs747691795 Chr.11: 47157902-47157902 on Build GRCh38
rs775835813 Chr.11: 47157844-47157844 on Build GRCh38
rs925016516 Chr.11: 47157851-47157851 on Build GRCh38
rs990962765 Chr.11: 47157857-47157857 on Build GRCh38
rs780962183 Chr.11: 47157804-47157804 on Build GRCh38
rs377553447 Chr.11: 47157960-47157960 on Build GRCh38
rs375778593 Chr.11: 47157874-47157874 on Build GRCh38
rs768457220 Chr.11: 47157887-47157888 on Build GRCh38
rs763380822 Chr.11: 47157950-47157950 on Build GRCh38
rs530870886 Chr.11: 47157968-47157968 on Build GRCh38
rs748169935 Chr.11: 47157811-47157811 on Build GRCh38
rs371909151 Chr.11: 47157841-47157841 on Build GRCh38
rs766816971 Chr.11: 47157954-47157954 on Build GRCh38
rs775710255 Chr.11: 47157780-47157782 on Build GRCh38
rs369968562 Chr.11: 47157883-47157883 on Build GRCh38
rs148603147 Chr.11: 47157922-47157922 on Build GRCh38
rs761987922 Chr.11: 47157942-47157942 on Build GRCh38
rs374910309 Chr.11: 47157798-47157798 on Build GRCh38
rs774243608 Chr.11: 47157909-47157911 on Build GRCh38
rs1030239588 Chr.11: 47157970-47157970 on Build GRCh38
rs751894679 Chr.11: 47157959-47157959 on Build GRCh38
rs749298274 Chr.11: 47157820-47157820 on Build GRCh38
rs761475742 Chr.11: 47157911-47157912 on Build GRCh38
rs754277056 Chr.11: 47157872-47157872 on Build GRCh38
rs575932294 Chr.11: 47157758-47157758 on Build GRCh38
rs977747690 Chr.11: 47157980-47157980 on Build GRCh38
rs769656828 Chr.11: 47157903-47157903 on Build GRCh38
rs765827421 Chr.11: 47157944-47157944 on Build GRCh38
rs375426513 Chr.11: 47157879-47157879 on Build GRCh38
rs900348331 Chr.11: 47157958-47157958 on Build GRCh38
rs768279071 Chr.11: 47157803-47157803 on Build GRCh38
rs560223007 Chr.11: 47157907-47157907 on Build GRCh38
rs750838821 Chr.11: 47157792-47157792 on Build GRCh38
rs143884410 Chr.11: 47157921-47157921 on Build GRCh38
rs758892753 Chr.11: 47157884-47157884 on Build GRCh38
rs371449285 Chr.11: 47157901-47157901 on Build GRCh38
rs774371269 Chr.11: 47157867-47157869 on Build GRCh38
rs184048914 Chr.11: 47157948-47157948 on Build GRCh38
rs760820668 Chr.11: 47157771-47157771 on Build GRCh38
rs762738071 Chr.11: 47157840-47157840 on Build GRCh38
rs752141042 Chr.11: 47157974-47157974 on Build GRCh38
rs776731932 Chr.11: 47157929-47157929 on Build GRCh38
rs757664412 Chr.11: 47157782-47157782 on Build GRCh38
rs966651339 Chr.11: 47157971-47157971 on Build GRCh38
rs190300633 Chr.11: 47157773-47157773 on Build GRCh38
rs966347646 Chr.11: 47157835-47157835 on Build GRCh38
rs186900565 Chr.11: 47157912-47157912 on Build GRCh38
rs754431846 Chr.11: 47157780-47157780 on Build GRCh38
rs758083253 Chr.11: 47157796-47157796 on Build GRCh38
rs200358227 Chr.11: 47157817-47157817 on Build GRCh38
rs775540324 Chr.11: 47157920-47157920 on Build GRCh38
rs764273209 Chr.11: 47157856-47157856 on Build GRCh38
rs888528147 Chr.11: 47157939-47157939 on Build GRCh38
rs760409759 Chr.11: 47157870-47157870 on Build GRCh38
rs34768698 Chr.11: 47157972-47157973 on Build GRCh38
rs770776665 Chr.11: 47157918-47157918 on Build GRCh38
rs201794825 Chr.11: 47157972-47157972 on Build GRCh38
rs200619017 Chr.11: 47157905-47157905 on Build GRCh38
rs772953685 Chr.11: 47157820-47157820 on Build GRCh38
rs560976157 Chr.11: 47157800-47157800 on Build GRCh38
rs149530929 Chr.11: 47157852-47157852 on Build GRCh38

View Full Product Details