Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23713028-23713301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT9
Entrez Gene ID
23597
Gene Name
acyl-CoA thioesterase 9
Gene Aliases
ACATE2, CGI-16, MT-ACT48, MTACT48
UniGene
Hs.298885
Gene Chromosome Location
Chr.X: 23701055-23743290 on Build GRCh38
Transcripts
NM_001033583.2 NM_001037171.1 XM_005274472.2

SNP Detail

SNP ID Chromosome Location
rs1023093832 Chr.X: 23713263-23713263 on Build GRCh38
rs762640576 Chr.X: 23713084-23713084 on Build GRCh38
rs750280804 Chr.X: 23713262-23713263 on Build GRCh38
rs770845318 Chr.X: 23713236-23713237 on Build GRCh38
rs372387863 Chr.X: 23713111-23713111 on Build GRCh38
rs186269629 Chr.X: 23713193-23713193 on Build GRCh38
rs778805402 Chr.X: 23713261-23713261 on Build GRCh38
rs766006923 Chr.X: 23713086-23713086 on Build GRCh38
rs888996533 Chr.X: 23713145-23713145 on Build GRCh38
rs1018918932 Chr.X: 23713157-23713157 on Build GRCh38
rs1007499127 Chr.X: 23713146-23713146 on Build GRCh38
rs752666995 Chr.X: 23713137-23713137 on Build GRCh38
rs993536719 Chr.X: 23713223-23713223 on Build GRCh38
rs768054940 Chr.X: 23713082-23713082 on Build GRCh38
rs746100629 Chr.X: 23713186-23713186 on Build GRCh38
rs146371808 Chr.X: 23713088-23713088 on Build GRCh38
rs777192782 Chr.X: 23713233-23713233 on Build GRCh38
rs762354437 Chr.X: 23713234-23713234 on Build GRCh38
rs201481463 Chr.X: 23713218-23713218 on Build GRCh38
rs369658267 Chr.X: 23713246-23713246 on Build GRCh38
rs1010734359 Chr.X: 23713108-23713108 on Build GRCh38
rs773573037 Chr.X: 23713240-23713240 on Build GRCh38
rs768711886 Chr.X: 23713226-23713226 on Build GRCh38
rs759173503 Chr.X: 23713256-23713256 on Build GRCh38
rs1052594518 Chr.X: 23713072-23713072 on Build GRCh38
rs970262414 Chr.X: 23713270-23713270 on Build GRCh38
rs148179443 Chr.X: 23713204-23713204 on Build GRCh38
rs754969124 Chr.X: 23713110-23713110 on Build GRCh38
rs1029527317 Chr.X: 23713265-23713265 on Build GRCh38
rs766205979 Chr.X: 23713236-23713236 on Build GRCh38
rs777602199 Chr.X: 23713179-23713179 on Build GRCh38
rs376691316 Chr.X: 23713178-23713178 on Build GRCh38
rs757517622 Chr.X: 23713252-23713252 on Build GRCh38

View Full Product Details