Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 70698450-70698711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
UniGene
Hs.21631
Gene Chromosome Location
Chr.7: 69598475-70793068 on Build GRCh38
Transcripts
NM_001127231.2 NM_015570.3 XM_011516010.2 XM_011516011.2 XM_011516012.2 XM_011516013.2 XM_011516014.2 XM_011516017.2 XM_011516018.2 XM_017011951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427696 c.718T>C p.F240L Chr.7: 70698596-70698596 on Build GRCh38
6608914 c.691-8delT p.? Chr.7: 70698561-70698561 on Build GRCh38
4420779 c.695C>T p.S232L Chr.7: 70698573-70698573 on Build GRCh38
237033 c.696A>G p.S232S Chr.7: 70698574-70698574 on Build GRCh38
7160230 c.704G>T p.S235I Chr.7: 70698582-70698582 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188901626 Chr.7: 70698529-70698529 on Build GRCh38
rs552747625 Chr.7: 70698470-70698470 on Build GRCh38
rs766612632 Chr.7: 70698532-70698532 on Build GRCh38
rs182043086 Chr.7: 70698601-70698601 on Build GRCh38
rs906688587 Chr.7: 70698675-70698675 on Build GRCh38
rs753881389 Chr.7: 70698535-70698535 on Build GRCh38
rs943986361 Chr.7: 70698480-70698480 on Build GRCh38
rs745724085 Chr.7: 70698581-70698581 on Build GRCh38
rs186099530 Chr.7: 70698469-70698469 on Build GRCh38
rs541389915 Chr.7: 70698560-70698560 on Build GRCh38
rs754615187 Chr.7: 70698543-70698544 on Build GRCh38
rs750865541 Chr.7: 70698528-70698528 on Build GRCh38
rs749219477 Chr.7: 70698552-70698552 on Build GRCh38
rs765630201 Chr.7: 70698524-70698524 on Build GRCh38
rs778803723 Chr.7: 70698559-70698559 on Build GRCh38
rs780266135 Chr.7: 70698589-70698589 on Build GRCh38
rs777288468 Chr.7: 70698549-70698549 on Build GRCh38
rs777595635 Chr.7: 70698513-70698514 on Build GRCh38
rs767347796 Chr.7: 70698530-70698530 on Build GRCh38
rs923667325 Chr.7: 70698505-70698505 on Build GRCh38
rs755594563 Chr.7: 70698548-70698548 on Build GRCh38
rs1042274547 Chr.7: 70698502-70698502 on Build GRCh38
rs753506410 Chr.7: 70698544-70698544 on Build GRCh38
rs999342297 Chr.7: 70698495-70698495 on Build GRCh38
rs757262379 Chr.7: 70698557-70698557 on Build GRCh38
rs768645278 Chr.7: 70698611-70698611 on Build GRCh38
rs370439387 Chr.7: 70698637-70698637 on Build GRCh38
rs921922598 Chr.7: 70698603-70698603 on Build GRCh38
rs771786097 Chr.7: 70698583-70698583 on Build GRCh38
rs776811681 Chr.7: 70698616-70698616 on Build GRCh38

View Full Product Details