Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1436560-1436797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC154
Entrez Gene ID
645811
Gene Name
coiled-coil domain containing 154
Gene Aliases
C16orf29
UniGene
Hs.355232
Gene Chromosome Location
Chr.16: 1434388-1444489 on Build GRCh38
Transcripts
NM_001143980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
396931 c.1342G>C p.E448Q Chr.16: 1436787-1436787 on Build GRCh38
2144032 c.1344G>A p.E448E Chr.16: 1436785-1436785 on Build GRCh38
2144031 c.1356G>A p.L452L Chr.16: 1436773-1436773 on Build GRCh38
3817235 c.1337G>A p.G446D Chr.16: 1436792-1436792 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766828357 Chr.16: 1436731-1436731 on Build GRCh38
rs759709659 Chr.16: 1436647-1436648 on Build GRCh38
rs899008985 Chr.16: 1436664-1436664 on Build GRCh38
rs958680092 Chr.16: 1436758-1436758 on Build GRCh38
rs538546813 Chr.16: 1436739-1436739 on Build GRCh38
rs765564071 Chr.16: 1436753-1436753 on Build GRCh38
rs758266311 Chr.16: 1436772-1436772 on Build GRCh38
rs76876166 Chr.16: 1436597-1436597 on Build GRCh38
rs968149139 Chr.16: 1436720-1436720 on Build GRCh38
rs888779016 Chr.16: 1436636-1436636 on Build GRCh38
rs916823439 Chr.16: 1436773-1436773 on Build GRCh38
rs906423935 Chr.16: 1436725-1436725 on Build GRCh38
rs991617593 Chr.16: 1436776-1436776 on Build GRCh38
rs375262245 Chr.16: 1436686-1436686 on Build GRCh38
rs759469892 Chr.16: 1436745-1436745 on Build GRCh38
rs978554617 Chr.16: 1436729-1436729 on Build GRCh38
rs1051396816 Chr.16: 1436620-1436620 on Build GRCh38
rs574311061 Chr.16: 1436754-1436754 on Build GRCh38
rs886076421 Chr.16: 1436625-1436625 on Build GRCh38
rs947529521 Chr.16: 1436585-1436585 on Build GRCh38
rs112015572 Chr.16: 1436699-1436699 on Build GRCh38
rs914770937 Chr.16: 1436584-1436584 on Build GRCh38
rs757251838 Chr.16: 1436615-1436615 on Build GRCh38
rs995020439 Chr.16: 1436605-1436605 on Build GRCh38
rs549594902 Chr.16: 1436669-1436669 on Build GRCh38
rs964628949 Chr.16: 1436667-1436667 on Build GRCh38
rs1038977144 Chr.16: 1436591-1436591 on Build GRCh38
rs1007351130 Chr.16: 1436643-1436643 on Build GRCh38
rs1008585185 Chr.16: 1436647-1436647 on Build GRCh38
rs1037841343 Chr.16: 1436645-1436645 on Build GRCh38
rs748613307 Chr.16: 1436666-1436666 on Build GRCh38

View Full Product Details