Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64270605-64270830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf56
Entrez Gene ID
115749
Gene Name
chromosome 12 open reading frame 56
Gene Aliases
-
UniGene
Hs.535190
Gene Chromosome Location
Chr.12: 64264762-64390580 on Build GRCh38
Transcripts
NM_001099676.2 NM_001170633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246427 c.1171A>C p.S391R Chr.12: 64270648-64270648 on Build GRCh38
405184 c.1153delG p.A385fs*6 Chr.12: 64270666-64270666 on Build GRCh38
4853398 c.1162C>G p.Q388E Chr.12: 64270657-64270657 on Build GRCh38
4044145 c.1136T>C p.V379A Chr.12: 64270683-64270683 on Build GRCh38
7156045 c.1167G>C p.L389L Chr.12: 64270652-64270652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556576427 Chr.12: 64270687-64270687 on Build GRCh38
rs1036232011 Chr.12: 64270648-64270648 on Build GRCh38
rs765932215 Chr.12: 64270643-64270643 on Build GRCh38
rs531778585 Chr.12: 64270794-64270794 on Build GRCh38
rs769126491 Chr.12: 64270736-64270736 on Build GRCh38
rs777178255 Chr.12: 64270740-64270740 on Build GRCh38
rs4499086 Chr.12: 64270696-64270696 on Build GRCh38
rs778998818 Chr.12: 64270710-64270710 on Build GRCh38
rs757176019 Chr.12: 64270707-64270707 on Build GRCh38
rs766884022 Chr.12: 64270764-64270764 on Build GRCh38
rs748503592 Chr.12: 64270752-64270752 on Build GRCh38
rs1046462615 Chr.12: 64270784-64270784 on Build GRCh38
rs140819146 Chr.12: 64270788-64270788 on Build GRCh38
rs35794015 Chr.12: 64270754-64270754 on Build GRCh38
rs766293192 Chr.12: 64270736-64270736 on Build GRCh38
rs767070566 Chr.12: 64270681-64270681 on Build GRCh38
rs201212935 Chr.12: 64270640-64270640 on Build GRCh38
rs780212543 Chr.12: 64270724-64270724 on Build GRCh38
rs747129739 Chr.12: 64270735-64270735 on Build GRCh38
rs758360036 Chr.12: 64270719-64270719 on Build GRCh38
rs751288206 Chr.12: 64270654-64270654 on Build GRCh38
rs758790191 Chr.12: 64270680-64270680 on Build GRCh38
rs746113930 Chr.12: 64270713-64270713 on Build GRCh38
rs373187432 Chr.12: 64270668-64270668 on Build GRCh38
rs559418283 Chr.12: 64270749-64270750 on Build GRCh38
rs756154476 Chr.12: 64270684-64270684 on Build GRCh38
rs1028838845 Chr.12: 64270699-64270699 on Build GRCh38
rs977544272 Chr.12: 64270797-64270797 on Build GRCh38
rs753664992 Chr.12: 64270690-64270690 on Build GRCh38

View Full Product Details