Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78239936-78240197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3382573 c.283C>T p.R95C Chr.9: 78240052-78240052 on Build GRCh38
7317441 c.356C>G p.S119* Chr.9: 78240125-78240125 on Build GRCh38
462376 c.418C>T p.L140L Chr.9: 78240187-78240187 on Build GRCh38
3218479 c.292G>T p.A98S Chr.9: 78240061-78240061 on Build GRCh38
1463131 c.405_425del21 p.D136_K142delDLTILAK Chr.9: 78240174-78240194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748644027 Chr.9: 78240125-78240125 on Build GRCh38
rs566305436 Chr.9: 78239982-78239982 on Build GRCh38
rs368318125 Chr.9: 78240063-78240063 on Build GRCh38
rs778505496 Chr.9: 78239985-78239985 on Build GRCh38
rs1042726781 Chr.9: 78240092-78240092 on Build GRCh38
rs34158345 Chr.9: 78240011-78240012 on Build GRCh38
rs763213647 Chr.9: 78240052-78240052 on Build GRCh38
rs367908635 Chr.9: 78240148-78240148 on Build GRCh38
rs770524086 Chr.9: 78239979-78239979 on Build GRCh38
rs367709973 Chr.9: 78240029-78240029 on Build GRCh38
rs766696825 Chr.9: 78240150-78240150 on Build GRCh38
rs773451169 Chr.9: 78240044-78240044 on Build GRCh38
rs760356786 Chr.9: 78240161-78240161 on Build GRCh38
rs747401879 Chr.9: 78240009-78240009 on Build GRCh38
rs199750679 Chr.9: 78240151-78240151 on Build GRCh38
rs747318619 Chr.9: 78240088-78240088 on Build GRCh38
rs769035638 Chr.9: 78240010-78240010 on Build GRCh38
rs775771977 Chr.9: 78240004-78240004 on Build GRCh38
rs779337546 Chr.9: 78240145-78240146 on Build GRCh38
rs773555625 Chr.9: 78240026-78240026 on Build GRCh38
rs776895025 Chr.9: 78240110-78240110 on Build GRCh38
rs749817667 Chr.9: 78240110-78240111 on Build GRCh38
rs200336329 Chr.9: 78239992-78239992 on Build GRCh38
rs372000838 Chr.9: 78240087-78240087 on Build GRCh38
rs777249352 Chr.9: 78240011-78240011 on Build GRCh38
rs868342001 Chr.9: 78240109-78240110 on Build GRCh38
rs745481521 Chr.9: 78239988-78239988 on Build GRCh38
rs199934742 Chr.9: 78240140-78240140 on Build GRCh38
rs757976688 Chr.9: 78240083-78240083 on Build GRCh38
rs574396568 Chr.9: 78240064-78240064 on Build GRCh38
rs371010271 Chr.9: 78240053-78240053 on Build GRCh38
rs768971127 Chr.9: 78240116-78240121 on Build GRCh38
rs748735168 Chr.9: 78239976-78239976 on Build GRCh38
rs375569426 Chr.9: 78240062-78240062 on Build GRCh38
rs778344131 Chr.9: 78240080-78240080 on Build GRCh38
rs887687984 Chr.9: 78240065-78240065 on Build GRCh38
rs754793883 Chr.9: 78239989-78239989 on Build GRCh38
rs770440873 Chr.9: 78240128-78240128 on Build GRCh38
rs780517670 Chr.9: 78240066-78240067 on Build GRCh38
rs778874630 Chr.9: 78240103-78240103 on Build GRCh38
rs745393868 Chr.9: 78240082-78240082 on Build GRCh38
rs757172721 Chr.9: 78240078-78240078 on Build GRCh38

View Full Product Details