Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207756859-207757108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD46
Entrez Gene ID
4179
Gene Name
CD46 molecule
Gene Aliases
AHUS2, MCP, MIC10, TLX, TRA2.10
UniGene
Hs.510402
Gene Chromosome Location
Chr.1: 207752038-207795516 on Build GRCh38
Transcripts
NM_002389.4 NM_153826.3 NM_172350.2 NM_172351.2 NM_172352.2 NM_172353.2 NM_172359.2 NM_172361.2 XM_011509563.1 XM_011509564.1 XM_017001308.1 XM_017001309.1 XM_017001310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551144 c.104G>A p.C35Y Chr.1: 207757020-207757020 on Build GRCh38
3671611 c.175C>A p.R59R Chr.1: 207757091-207757091 on Build GRCh38
3803726 c.138C>T p.L46L Chr.1: 207757054-207757054 on Build GRCh38
3966231 c.121T>C p.F41L Chr.1: 207757037-207757037 on Build GRCh38
1320521 c.98A>G p.D33G Chr.1: 207757014-207757014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777524358 Chr.1: 207757008-207757008 on Build GRCh38
rs573909363 Chr.1: 207756886-207756886 on Build GRCh38
rs564910868 Chr.1: 207757046-207757046 on Build GRCh38
rs758670382 Chr.1: 207756961-207756961 on Build GRCh38
rs753141463 Chr.1: 207757076-207757076 on Build GRCh38
rs987716412 Chr.1: 207756919-207756920 on Build GRCh38
rs943330063 Chr.1: 207756947-207756947 on Build GRCh38
rs978529132 Chr.1: 207756889-207756889 on Build GRCh38
rs757181191 Chr.1: 207757064-207757065 on Build GRCh38
rs113181165 Chr.1: 207756912-207756912 on Build GRCh38
rs531437382 Chr.1: 207756984-207756984 on Build GRCh38
rs753809279 Chr.1: 207757061-207757064 on Build GRCh38
rs191877084 Chr.1: 207756893-207756893 on Build GRCh38
rs755559270 Chr.1: 207757043-207757043 on Build GRCh38
rs766600746 Chr.1: 207757026-207757026 on Build GRCh38
rs763780170 Chr.1: 207756996-207756996 on Build GRCh38
rs562619993 Chr.1: 207756943-207756943 on Build GRCh38
rs753924720 Chr.1: 207757034-207757034 on Build GRCh38
rs544735882 Chr.1: 207757018-207757018 on Build GRCh38
rs121909591 Chr.1: 207757020-207757020 on Build GRCh38
rs911775849 Chr.1: 207756922-207756922 on Build GRCh38
rs1031854381 Chr.1: 207756893-207756894 on Build GRCh38

View Full Product Details