Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43191362-43191587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNDBP1
Entrez Gene ID
23582
Gene Name
cyclin D1 binding protein 1
Gene Aliases
DIP1, GCIP, HHM
UniGene
Hs.36794
Gene Chromosome Location
Chr.15: 43185118-43197177 on Build GRCh38
Transcripts
NM_012142.4 NR_027513.2 NR_027514.2 NR_045998.1 NR_045999.1 XM_006720448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
961896 c.748G>A p.A250T Chr.15: 43191563-43191563 on Build GRCh38
961895 c.719T>G p.L240R Chr.15: 43191534-43191534 on Build GRCh38
6622982 c.660T>C p.D220D Chr.15: 43191475-43191475 on Build GRCh38
4054772 c.615G>T p.L205F Chr.15: 43191430-43191430 on Build GRCh38
6622981 c.627G>A p.E209E Chr.15: 43191442-43191442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762801668 Chr.15: 43191521-43191521 on Build GRCh38
rs778288079 Chr.15: 43191450-43191450 on Build GRCh38
rs182468887 Chr.15: 43191392-43191392 on Build GRCh38
rs565667051 Chr.15: 43191453-43191453 on Build GRCh38
rs753079979 Chr.15: 43191543-43191543 on Build GRCh38
rs925458780 Chr.15: 43191471-43191471 on Build GRCh38
rs772212732 Chr.15: 43191482-43191482 on Build GRCh38
rs771268690 Chr.15: 43191472-43191472 on Build GRCh38
rs750871228 Chr.15: 43191548-43191548 on Build GRCh38
rs754554121 Chr.15: 43191432-43191432 on Build GRCh38
rs373552340 Chr.15: 43191436-43191436 on Build GRCh38
rs143142653 Chr.15: 43191523-43191523 on Build GRCh38
rs145310962 Chr.15: 43191399-43191399 on Build GRCh38
rs770244627 Chr.15: 43191440-43191440 on Build GRCh38
rs754399432 Chr.15: 43191529-43191529 on Build GRCh38
rs745783242 Chr.15: 43191558-43191559 on Build GRCh38
rs774705009 Chr.15: 43191476-43191476 on Build GRCh38
rs764950508 Chr.15: 43191511-43191511 on Build GRCh38
rs779738641 Chr.15: 43191412-43191412 on Build GRCh38
rs201609265 Chr.15: 43191524-43191524 on Build GRCh38
rs748713511 Chr.15: 43191439-43191439 on Build GRCh38
rs115809459 Chr.15: 43191553-43191553 on Build GRCh38
rs758060193 Chr.15: 43191404-43191404 on Build GRCh38
rs376549003 Chr.15: 43191519-43191519 on Build GRCh38
rs757739759 Chr.15: 43191554-43191554 on Build GRCh38
rs749468282 Chr.15: 43191459-43191459 on Build GRCh38
rs751027223 Chr.15: 43191418-43191418 on Build GRCh38
rs761811013 Chr.15: 43191508-43191508 on Build GRCh38
rs762040761 Chr.15: 43191516-43191516 on Build GRCh38
rs528672559 Chr.15: 43191552-43191552 on Build GRCh38
rs780786932 Chr.15: 43191535-43191535 on Build GRCh38
rs149102527 Chr.15: 43191501-43191501 on Build GRCh38
rs778917726 Chr.15: 43191475-43191479 on Build GRCh38
rs147637019 Chr.15: 43191402-43191402 on Build GRCh38
rs867909765 Chr.15: 43191449-43191449 on Build GRCh38
rs759506354 Chr.15: 43191481-43191481 on Build GRCh38
rs917688383 Chr.15: 43191517-43191517 on Build GRCh38
rs966948834 Chr.15: 43191429-43191429 on Build GRCh38

View Full Product Details