Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58211219-58211450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
UniGene
Hs.509451
Gene Chromosome Location
Chr.14: 58200115-58235636 on Build GRCh38
Transcripts
NM_018477.2 XM_011536960.1 XM_011536961.1 XM_011536962.2 XM_011536963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4051435 c.420T>A p.D140E Chr.14: 58211369-58211369 on Build GRCh38
698276 c.441G>T p.L147L Chr.14: 58211390-58211390 on Build GRCh38
4051434 c.385C>T p.L129L Chr.14: 58211334-58211334 on Build GRCh38
6593071 c.367A>C p.S123R Chr.14: 58211316-58211316 on Build GRCh38
7122026 c.390G>A p.T130T Chr.14: 58211339-58211339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762003037 Chr.14: 58211297-58211297 on Build GRCh38
rs17832891 Chr.14: 58211339-58211339 on Build GRCh38
rs781743036 Chr.14: 58211393-58211393 on Build GRCh38
rs773598407 Chr.14: 58211304-58211304 on Build GRCh38
rs371671757 Chr.14: 58211313-58211313 on Build GRCh38
rs754722393 Chr.14: 58211338-58211338 on Build GRCh38
rs755685616 Chr.14: 58211346-58211346 on Build GRCh38
rs756839879 Chr.14: 58211371-58211371 on Build GRCh38
rs769274206 Chr.14: 58211275-58211275 on Build GRCh38
rs772452142 Chr.14: 58211261-58211261 on Build GRCh38
rs778534247 Chr.14: 58211377-58211377 on Build GRCh38
rs766288805 Chr.14: 58211316-58211316 on Build GRCh38
rs747433920 Chr.14: 58211378-58211378 on Build GRCh38
rs748904123 Chr.14: 58211260-58211260 on Build GRCh38
rs1001407339 Chr.14: 58211280-58211280 on Build GRCh38
rs139523686 Chr.14: 58211324-58211324 on Build GRCh38
rs777101475 Chr.14: 58211358-58211358 on Build GRCh38
rs776991031 Chr.14: 58211281-58211281 on Build GRCh38
rs145464377 Chr.14: 58211400-58211400 on Build GRCh38
rs765199716 Chr.14: 58211301-58211301 on Build GRCh38
rs748563352 Chr.14: 58211399-58211399 on Build GRCh38
rs747520692 Chr.14: 58211271-58211271 on Build GRCh38
rs768882121 Chr.14: 58211390-58211390 on Build GRCh38
rs748955462 Chr.14: 58211361-58211361 on Build GRCh38
rs773367161 Chr.14: 58211402-58211402 on Build GRCh38
rs866990657 Chr.14: 58211389-58211389 on Build GRCh38
rs775776354 Chr.14: 58211268-58211268 on Build GRCh38
rs777526452 Chr.14: 58211259-58211259 on Build GRCh38

View Full Product Details