Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129615285-129615546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
-
UniGene
Hs.124223
Gene Chromosome Location
Chr.9: 129612225-129622275 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3328414 c.832G>A p.E278K Chr.9: 129615532-129615532 on Build GRCh38
1105850 c.820C>T p.R274* Chr.9: 129615544-129615544 on Build GRCh38
1755989 c.851C>T p.T284M Chr.9: 129615513-129615513 on Build GRCh38
5410519 c.844G>A p.D282N Chr.9: 129615520-129615520 on Build GRCh38
5472673 c.825C>T p.F275F Chr.9: 129615539-129615539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759914443 Chr.9: 129615513-129615513 on Build GRCh38
rs562595067 Chr.9: 129615396-129615396 on Build GRCh38
rs376751991 Chr.9: 129615459-129615459 on Build GRCh38
rs766786474 Chr.9: 129615452-129615452 on Build GRCh38
rs2275898 Chr.9: 129615345-129615345 on Build GRCh38
rs201594971 Chr.9: 129615461-129615461 on Build GRCh38
rs926811210 Chr.9: 129615515-129615515 on Build GRCh38
rs755502616 Chr.9: 129615485-129615486 on Build GRCh38
rs768174059 Chr.9: 129615445-129615445 on Build GRCh38
rs936809488 Chr.9: 129615413-129615413 on Build GRCh38
rs752795348 Chr.9: 129615409-129615409 on Build GRCh38
rs756742371 Chr.9: 129615471-129615471 on Build GRCh38
rs774340615 Chr.9: 129615434-129615434 on Build GRCh38
rs374809292 Chr.9: 129615495-129615495 on Build GRCh38
rs551125031 Chr.9: 129615485-129615485 on Build GRCh38
rs761137585 Chr.9: 129615450-129615450 on Build GRCh38
rs1054188080 Chr.9: 129615436-129615436 on Build GRCh38
rs146441613 Chr.9: 129615512-129615512 on Build GRCh38
rs770377536 Chr.9: 129615325-129615325 on Build GRCh38
rs547361651 Chr.9: 129615493-129615493 on Build GRCh38
rs904027164 Chr.9: 129615394-129615394 on Build GRCh38
rs140534012 Chr.9: 129615507-129615507 on Build GRCh38
rs751071708 Chr.9: 129615466-129615466 on Build GRCh38
rs918166 Chr.9: 129615449-129615449 on Build GRCh38
rs778411088 Chr.9: 129615496-129615496 on Build GRCh38
rs756724085 Chr.9: 129615528-129615528 on Build GRCh38
rs763685959 Chr.9: 129615514-129615514 on Build GRCh38
rs773863913 Chr.9: 129615503-129615503 on Build GRCh38
rs754252778 Chr.9: 129615458-129615458 on Build GRCh38
rs766024837 Chr.9: 129615307-129615307 on Build GRCh38
rs748547670 Chr.9: 129615528-129615532 on Build GRCh38
rs768796730 Chr.9: 129615492-129615492 on Build GRCh38
rs1028397406 Chr.9: 129615431-129615431 on Build GRCh38
rs1001325390 Chr.9: 129615403-129615403 on Build GRCh38
rs1041589194 Chr.9: 129615455-129615455 on Build GRCh38
rs772526424 Chr.9: 129615528-129615529 on Build GRCh38
rs781120632 Chr.9: 129615480-129615480 on Build GRCh38
rs117566704 Chr.9: 129615490-129615490 on Build GRCh38
rs779529919 Chr.9: 129615513-129615522 on Build GRCh38
rs768906452 Chr.9: 129615433-129615433 on Build GRCh38
rs200304993 Chr.9: 129615425-129615425 on Build GRCh38
rs369203583 Chr.9: 129615527-129615527 on Build GRCh38
rs896728775 Chr.9: 129615343-129615343 on Build GRCh38
rs112216725 Chr.9: 129615447-129615447 on Build GRCh38
rs149091556 Chr.9: 129615525-129615525 on Build GRCh38
rs997285786 Chr.9: 129615477-129615477 on Build GRCh38
rs561500325 Chr.9: 129615505-129615505 on Build GRCh38
rs776911121 Chr.9: 129615508-129615508 on Build GRCh38
rs937690853 Chr.9: 129615312-129615312 on Build GRCh38
rs779247408 Chr.9: 129615355-129615355 on Build GRCh38
rs375237794 Chr.9: 129615435-129615435 on Build GRCh38

View Full Product Details