Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150301950-150302175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSI
Entrez Gene ID
340075
Gene Name
arylsulfatase family member I
Gene Aliases
SPG66
UniGene
Hs.591252
Gene Chromosome Location
Chr.5: 150296346-150302962 on Build GRCh38
Transcripts
NM_001012301.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134557 c.222G>T p.L74L Chr.5: 150302152-150302152 on Build GRCh38
6535780 c.293G>C p.S98T Chr.5: 150302081-150302081 on Build GRCh38
318785 c.287C>G p.S96W Chr.5: 150302087-150302087 on Build GRCh38
1238234 c.223G>A p.D75N Chr.5: 150302151-150302151 on Build GRCh38
737232 c.289C>T p.R97W Chr.5: 150302085-150302085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982963651 Chr.5: 150302012-150302015 on Build GRCh38
rs932563113 Chr.5: 150302040-150302040 on Build GRCh38
rs370028777 Chr.5: 150302071-150302071 on Build GRCh38
rs772351600 Chr.5: 150302111-150302111 on Build GRCh38
rs925752168 Chr.5: 150302073-150302073 on Build GRCh38
rs143586885 Chr.5: 150302143-150302143 on Build GRCh38
rs992869381 Chr.5: 150302061-150302061 on Build GRCh38
rs199972086 Chr.5: 150302085-150302085 on Build GRCh38
rs769355277 Chr.5: 150302023-150302023 on Build GRCh38
rs941056812 Chr.5: 150302042-150302042 on Build GRCh38
rs199603906 Chr.5: 150302141-150302141 on Build GRCh38
rs73799004 Chr.5: 150301981-150301981 on Build GRCh38
rs764309148 Chr.5: 150302135-150302135 on Build GRCh38
rs960506981 Chr.5: 150302000-150302003 on Build GRCh38
rs774801277 Chr.5: 150302087-150302087 on Build GRCh38
rs139714700 Chr.5: 150302089-150302089 on Build GRCh38
rs1042963929 Chr.5: 150302067-150302067 on Build GRCh38
rs762816002 Chr.5: 150302139-150302139 on Build GRCh38
rs192076761 Chr.5: 150302012-150302012 on Build GRCh38
rs960053515 Chr.5: 150302014-150302014 on Build GRCh38
rs775632237 Chr.5: 150302126-150302126 on Build GRCh38
rs760802512 Chr.5: 150302131-150302131 on Build GRCh38
rs974014166 Chr.5: 150302082-150302082 on Build GRCh38
rs778251975 Chr.5: 150302033-150302033 on Build GRCh38
rs763003022 Chr.5: 150302142-150302142 on Build GRCh38
rs749835863 Chr.5: 150302066-150302066 on Build GRCh38

View Full Product Details