Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112742535-112742778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2L
Entrez Gene ID
150468
Gene Name
cytoskeleton associated protein 2 like
Gene Aliases
-
UniGene
Hs.434250
Gene Chromosome Location
Chr.2: 112736349-112764677 on Build GRCh38
Transcripts
NM_001304361.1 NM_152515.4 NR_130712.1 XM_011510666.2 XM_017005475.1

Gene Symbol
NT5DC4
Entrez Gene ID
284958
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
UniGene
Hs.589714
Gene Chromosome Location
Chr.2: 112717245-112742879 on Build GRCh38
Transcripts
NM_001304361.1 NM_152515.4 NR_130712.1 XM_011510666.2 XM_017005475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6730569 c.1759-6T>A p.? Chr.2: 112742775-112742775 on Build GRCh38
7170291 c.1771T>C p.L591L Chr.2: 112742757-112742757 on Build GRCh38
1683323 c.1794_1795insT p.L599fs*14 Chr.2: 112742733-112742734 on Build GRCh38
6087376 c.1821A>T p.E607D Chr.2: 112742707-112742707 on Build GRCh38
5500915 c.1818A>C p.T606T Chr.2: 112742710-112742710 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112440036 Chr.2: 112742673-112742673 on Build GRCh38
rs371885912 Chr.2: 112742753-112742753 on Build GRCh38
rs561549815 Chr.2: 112742754-112742754 on Build GRCh38
rs553122925 Chr.2: 112742639-112742639 on Build GRCh38
rs762572685 Chr.2: 112742658-112742658 on Build GRCh38
rs767401436 Chr.2: 112742694-112742694 on Build GRCh38
rs752569349 Chr.2: 112742698-112742698 on Build GRCh38
rs1029969186 Chr.2: 112742628-112742628 on Build GRCh38
rs757948812 Chr.2: 112742636-112742636 on Build GRCh38
rs746117908 Chr.2: 112742751-112742751 on Build GRCh38
rs375004681 Chr.2: 112742691-112742691 on Build GRCh38
rs756158416 Chr.2: 112742706-112742706 on Build GRCh38
rs1000390570 Chr.2: 112742568-112742568 on Build GRCh38
rs765968456 Chr.2: 112742680-112742680 on Build GRCh38
rs1052776433 Chr.2: 112742623-112742623 on Build GRCh38
rs138538433 Chr.2: 112742743-112742743 on Build GRCh38
rs891433603 Chr.2: 112742634-112742634 on Build GRCh38
rs753916738 Chr.2: 112742732-112742732 on Build GRCh38
rs757447326 Chr.2: 112742735-112742735 on Build GRCh38
rs369349558 Chr.2: 112742731-112742731 on Build GRCh38
rs764243826 Chr.2: 112742716-112742716 on Build GRCh38
rs774477317 Chr.2: 112742672-112742674 on Build GRCh38

View Full Product Details