Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 19838668-19838895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH18
Entrez Gene ID
1016
Gene Name
cadherin 18
Gene Aliases
CDH14, CDH14L, CDH24
UniGene
Hs.317632
Gene Chromosome Location
Chr.5: 19472930-20575902 on Build GRCh38
Transcripts
NM_001167667.1 NM_001291956.1 NM_001291957.1 NM_004934.3 XM_005248228.3 XM_006714435.3 XM_011513930.2 XM_017008924.1 XM_017008925.1 XM_017008926.1 XM_017008927.1 XM_017008928.1 XM_017008929.1 XM_017008930.1 XM_017008931.1 XM_017008932.1 XM_017008933.1 XM_017008934.1 XM_017008935.1 XM_017008936.1 XM_017008937.1 XM_017008938.1 XM_017008939.1 XM_017008940.1 XM_017008941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157333 c.188T>C p.L63S Chr.5: 19838799-19838799 on Build GRCh38
7153388 c.192A>T p.E64D Chr.5: 19838795-19838795 on Build GRCh38
6624749 c.117A>G p.K39K Chr.5: 19838870-19838870 on Build GRCh38
3947376 c.170G>T p.W57L Chr.5: 19838817-19838817 on Build GRCh38
7420166 c.169T>A p.W57R Chr.5: 19838818-19838818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055235253 Chr.5: 19838703-19838703 on Build GRCh38
rs961704288 Chr.5: 19838867-19838867 on Build GRCh38
rs907339902 Chr.5: 19838846-19838846 on Build GRCh38
rs772588846 Chr.5: 19838790-19838790 on Build GRCh38
rs1013601356 Chr.5: 19838768-19838768 on Build GRCh38
rs200906154 Chr.5: 19838834-19838834 on Build GRCh38
rs747474865 Chr.5: 19838833-19838833 on Build GRCh38
rs751391472 Chr.5: 19838866-19838866 on Build GRCh38
rs779360240 Chr.5: 19838786-19838786 on Build GRCh38
rs779949619 Chr.5: 19838712-19838712 on Build GRCh38
rs761770637 Chr.5: 19838841-19838841 on Build GRCh38
rs766419800 Chr.5: 19838852-19838852 on Build GRCh38
rs747594765 Chr.5: 19838754-19838754 on Build GRCh38
rs372817208 Chr.5: 19838851-19838851 on Build GRCh38
rs746332930 Chr.5: 19838788-19838788 on Build GRCh38
rs545255378 Chr.5: 19838730-19838730 on Build GRCh38
rs775765843 Chr.5: 19838821-19838821 on Build GRCh38
rs751417879 Chr.5: 19838728-19838728 on Build GRCh38
rs776963132 Chr.5: 19838839-19838839 on Build GRCh38
rs780027030 Chr.5: 19838745-19838745 on Build GRCh38
rs769392828 Chr.5: 19838774-19838774 on Build GRCh38
rs936340343 Chr.5: 19838700-19838700 on Build GRCh38
rs772234786 Chr.5: 19838729-19838730 on Build GRCh38
rs780960973 Chr.5: 19838753-19838753 on Build GRCh38
rs765370372 Chr.5: 19838844-19838844 on Build GRCh38

View Full Product Details