Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44408397-44408640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
UniGene
Hs.485471
Gene Chromosome Location
Chr.6: 44387514-44450424 on Build GRCh38
Transcripts
NM_001253.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670737 c.961C>T p.R321C Chr.6: 44408501-44408501 on Build GRCh38
6253843 c.1045G>T p.A349S Chr.6: 44408585-44408585 on Build GRCh38
1079789 c.1061G>A p.R354Q Chr.6: 44408601-44408601 on Build GRCh38
1079788 c.991A>G p.N331D Chr.6: 44408531-44408531 on Build GRCh38
5729648 c.1057C>T p.P353S Chr.6: 44408597-44408597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200986556 Chr.6: 44408572-44408572 on Build GRCh38
rs941493029 Chr.6: 44408502-44408502 on Build GRCh38
rs776484259 Chr.6: 44408581-44408581 on Build GRCh38
rs751834925 Chr.6: 44408505-44408505 on Build GRCh38
rs771602856 Chr.6: 44408552-44408552 on Build GRCh38
rs536765287 Chr.6: 44408520-44408520 on Build GRCh38
rs11552843 Chr.6: 44408498-44408498 on Build GRCh38
rs148664123 Chr.6: 44408600-44408600 on Build GRCh38
rs753603218 Chr.6: 44408453-44408453 on Build GRCh38
rs972004000 Chr.6: 44408579-44408579 on Build GRCh38
rs745430137 Chr.6: 44408538-44408538 on Build GRCh38
rs770590185 Chr.6: 44408421-44408421 on Build GRCh38
rs376004864 Chr.6: 44408565-44408565 on Build GRCh38
rs141567880 Chr.6: 44408455-44408455 on Build GRCh38
rs200890159 Chr.6: 44408547-44408547 on Build GRCh38
rs755405339 Chr.6: 44408506-44408506 on Build GRCh38
rs9381320 Chr.6: 44408535-44408535 on Build GRCh38
rs369777781 Chr.6: 44408482-44408482 on Build GRCh38
rs200087867 Chr.6: 44408469-44408469 on Build GRCh38
rs781506877 Chr.6: 44408512-44408512 on Build GRCh38
rs760568173 Chr.6: 44408461-44408461 on Build GRCh38
rs149525934 Chr.6: 44408475-44408475 on Build GRCh38
rs539044223 Chr.6: 44408614-44408614 on Build GRCh38
rs1006568146 Chr.6: 44408582-44408582 on Build GRCh38
rs1022052815 Chr.6: 44408491-44408491 on Build GRCh38
rs763145775 Chr.6: 44408500-44408500 on Build GRCh38
rs373916286 Chr.6: 44408487-44408487 on Build GRCh38
rs1051362293 Chr.6: 44408575-44408575 on Build GRCh38
rs771826078 Chr.6: 44408441-44408441 on Build GRCh38
rs891353395 Chr.6: 44408580-44408580 on Build GRCh38
rs930670024 Chr.6: 44408617-44408617 on Build GRCh38
rs748222402 Chr.6: 44408513-44408513 on Build GRCh38
rs372513973 Chr.6: 44408423-44408423 on Build GRCh38
rs778564165 Chr.6: 44408525-44408525 on Build GRCh38
rs892126229 Chr.6: 44408471-44408471 on Build GRCh38
rs750565964 Chr.6: 44408496-44408496 on Build GRCh38
rs377117589 Chr.6: 44408434-44408434 on Build GRCh38
rs1038853577 Chr.6: 44408564-44408564 on Build GRCh38
rs756763383 Chr.6: 44408523-44408523 on Build GRCh38
rs779796790 Chr.6: 44408563-44408563 on Build GRCh38
rs768509617 Chr.6: 44408568-44408568 on Build GRCh38
rs144083420 Chr.6: 44408501-44408501 on Build GRCh38

View Full Product Details