Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153725453-153725720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD1
Entrez Gene ID
215
Gene Name
ATP binding cassette subfamily D member 1
Gene Aliases
ABC42, ALD, ALDP, AMN
UniGene
Hs.159546
Gene Chromosome Location
Chr.X: 153724851-153744762 on Build GRCh38
Transcripts
NM_000033.3

SNP Detail

SNP ID Chromosome Location
rs1009106172 Chr.X: 153725633-153725633 on Build GRCh38
rs370602067 Chr.X: 153725532-153725532 on Build GRCh38
rs398123111 Chr.X: 153725672-153725672 on Build GRCh38
rs782107505 Chr.X: 153725692-153725692 on Build GRCh38
rs782221851 Chr.X: 153725520-153725520 on Build GRCh38
rs782394070 Chr.X: 153725541-153725541 on Build GRCh38
rs398123110 Chr.X: 153725612-153725612 on Build GRCh38
rs367799134 Chr.X: 153725658-153725658 on Build GRCh38
rs782661286 Chr.X: 153725649-153725649 on Build GRCh38
rs782121374 Chr.X: 153725624-153725624 on Build GRCh38
rs782677645 Chr.X: 153725531-153725531 on Build GRCh38
rs782359412 Chr.X: 153725652-153725652 on Build GRCh38
rs74315280 Chr.X: 153725661-153725661 on Build GRCh38
rs193922097 Chr.X: 153725687-153725687 on Build GRCh38
rs782574881 Chr.X: 153725522-153725522 on Build GRCh38
rs782628755 Chr.X: 153725515-153725515 on Build GRCh38
rs940741241 Chr.X: 153725500-153725500 on Build GRCh38
rs781954264 Chr.X: 153725560-153725560 on Build GRCh38
rs782499252 Chr.X: 153725648-153725648 on Build GRCh38
rs782816273 Chr.X: 153725599-153725599 on Build GRCh38
rs142950300 Chr.X: 153725585-153725585 on Build GRCh38
rs200660869 Chr.X: 153725524-153725524 on Build GRCh38
rs781883009 Chr.X: 153725491-153725491 on Build GRCh38
rs781846746 Chr.X: 153725621-153725621 on Build GRCh38
rs782138372 Chr.X: 153725592-153725592 on Build GRCh38
rs375790297 Chr.X: 153725587-153725587 on Build GRCh38
rs782458499 Chr.X: 153725636-153725636 on Build GRCh38
rs781797609 Chr.X: 153725625-153725625 on Build GRCh38
rs201979180 Chr.X: 153725540-153725540 on Build GRCh38
rs782098898 Chr.X: 153725566-153725566 on Build GRCh38
rs782377368 Chr.X: 153725701-153725701 on Build GRCh38
rs782537874 Chr.X: 153725497-153725497 on Build GRCh38
rs889405096 Chr.X: 153725563-153725563 on Build GRCh38
rs782197275 Chr.X: 153725680-153725680 on Build GRCh38
rs713993050 Chr.X: 153725519-153725520 on Build GRCh38

View Full Product Details