Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116760552-116760825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
UniGene
Hs.437341
Gene Chromosome Location
Chr.11: 116748170-116772998 on Build GRCh38
Transcripts
NM_001159736.1 NM_032725.3 XM_011543035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274183 c.1229G>A p.R410Q Chr.11: 116760760-116760760 on Build GRCh38
4666431 c.1228C>T p.R410* Chr.11: 116760761-116760761 on Build GRCh38
3675985 c.1218G>C p.L406L Chr.11: 116760771-116760771 on Build GRCh38
3356162 c.1254+2T>A p.? Chr.11: 116760733-116760733 on Build GRCh38
1246325 c.1254+4A>G p.? Chr.11: 116760731-116760731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868376033 Chr.11: 116760729-116760729 on Build GRCh38
rs758736430 Chr.11: 116760771-116760771 on Build GRCh38
rs964898451 Chr.11: 116760620-116760620 on Build GRCh38
rs61730763 Chr.11: 116760766-116760766 on Build GRCh38
rs3741301 Chr.11: 116760675-116760675 on Build GRCh38
rs748247607 Chr.11: 116760805-116760805 on Build GRCh38
rs755516486 Chr.11: 116760793-116760793 on Build GRCh38
rs764862821 Chr.11: 116760765-116760765 on Build GRCh38
rs753401362 Chr.11: 116760760-116760760 on Build GRCh38
rs990093776 Chr.11: 116760622-116760622 on Build GRCh38
rs560234515 Chr.11: 116760618-116760618 on Build GRCh38
rs373740790 Chr.11: 116760702-116760702 on Build GRCh38
rs139212887 Chr.11: 116760753-116760753 on Build GRCh38
rs760185279 Chr.11: 116760603-116760603 on Build GRCh38
rs781078672 Chr.11: 116760794-116760794 on Build GRCh38
rs772033303 Chr.11: 116760707-116760707 on Build GRCh38
rs781366978 Chr.11: 116760648-116760648 on Build GRCh38
rs775241181 Chr.11: 116760718-116760718 on Build GRCh38
rs780421872 Chr.11: 116760777-116760777 on Build GRCh38
rs554805018 Chr.11: 116760643-116760643 on Build GRCh38
rs370077873 Chr.11: 116760701-116760701 on Build GRCh38
rs770154878 Chr.11: 116760688-116760688 on Build GRCh38
rs374514869 Chr.11: 116760644-116760644 on Build GRCh38
rs1055036676 Chr.11: 116760609-116760609 on Build GRCh38
rs190341215 Chr.11: 116760706-116760706 on Build GRCh38
rs751974437 Chr.11: 116760782-116760782 on Build GRCh38
rs1003222666 Chr.11: 116760710-116760710 on Build GRCh38
rs945648072 Chr.11: 116760669-116760669 on Build GRCh38
rs760697657 Chr.11: 116760747-116760747 on Build GRCh38

View Full Product Details