Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195568693-195568950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOD
Entrez Gene ID
347
Gene Name
apolipoprotein D
Gene Aliases
-
UniGene
Hs.522555
Gene Chromosome Location
Chr.3: 195568702-195584205 on Build GRCh38
Transcripts
NM_001647.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190499 c.521T>A p.V174D Chr.3: 195568949-195568949 on Build GRCh38
7302785 c.533C>T p.T178M Chr.3: 195568937-195568937 on Build GRCh38
6602422 c.523A>G p.K175E Chr.3: 195568947-195568947 on Build GRCh38
730625 c.522C>A p.V174V Chr.3: 195568948-195568948 on Build GRCh38
1245242 c.523A>C p.K175Q Chr.3: 195568947-195568947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049096862 Chr.3: 195568764-195568764 on Build GRCh38
rs142189206 Chr.3: 195568837-195568838 on Build GRCh38
rs1014384782 Chr.3: 195568827-195568827 on Build GRCh38
rs369497400 Chr.3: 195568833-195568834 on Build GRCh38
rs111354824 Chr.3: 195568837-195568837 on Build GRCh38
rs377294046 Chr.3: 195568904-195568904 on Build GRCh38
rs375215213 Chr.3: 195568839-195568840 on Build GRCh38
rs757306351 Chr.3: 195568861-195568861 on Build GRCh38
rs746682027 Chr.3: 195568895-195568895 on Build GRCh38
rs542624395 Chr.3: 195568814-195568814 on Build GRCh38
rs4651 Chr.3: 195568834-195568834 on Build GRCh38
rs367839491 Chr.3: 195568838-195568839 on Build GRCh38
rs554960514 Chr.3: 195568832-195568832 on Build GRCh38
rs200510277 Chr.3: 195568903-195568903 on Build GRCh38
rs932782307 Chr.3: 195568870-195568870 on Build GRCh38
rs201398382 Chr.3: 195568831-195568831 on Build GRCh38
rs79725750 Chr.3: 195568734-195568734 on Build GRCh38
rs1043060634 Chr.3: 195568769-195568769 on Build GRCh38
rs753172328 Chr.3: 195568721-195568721 on Build GRCh38
rs878962236 Chr.3: 195568842-195568842 on Build GRCh38
rs924390379 Chr.3: 195568846-195568846 on Build GRCh38
rs150401209 Chr.3: 195568911-195568911 on Build GRCh38
rs1015918248 Chr.3: 195568835-195568835 on Build GRCh38
rs8306 Chr.3: 195568809-195568809 on Build GRCh38
rs7659 Chr.3: 195568790-195568790 on Build GRCh38
rs964641049 Chr.3: 195568881-195568881 on Build GRCh38
rs993174403 Chr.3: 195568838-195568838 on Build GRCh38
rs137988905 Chr.3: 195568808-195568808 on Build GRCh38
rs925980803 Chr.3: 195568778-195568778 on Build GRCh38
rs779437607 Chr.3: 195568862-195568862 on Build GRCh38
rs200334618 Chr.3: 195568852-195568852 on Build GRCh38
rs772547403 Chr.3: 195568866-195568866 on Build GRCh38
rs71180934 Chr.3: 195568840-195568841 on Build GRCh38
rs572944911 Chr.3: 195568836-195568836 on Build GRCh38
rs201525248 Chr.3: 195568843-195568843 on Build GRCh38
rs746372011 Chr.3: 195568863-195568863 on Build GRCh38
rs67866189 Chr.3: 195568843-195568844 on Build GRCh38
rs532947188 Chr.3: 195568909-195568909 on Build GRCh38
rs780229357 Chr.3: 195568872-195568872 on Build GRCh38
rs202214202 Chr.3: 195568853-195568853 on Build GRCh38
rs190533362 Chr.3: 195568751-195568751 on Build GRCh38
rs773348727 Chr.3: 195568923-195568923 on Build GRCh38
rs146445395 Chr.3: 195568831-195568832 on Build GRCh38
rs1026043520 Chr.3: 195568839-195568839 on Build GRCh38
rs778372877 Chr.3: 195568770-195568770 on Build GRCh38

View Full Product Details