Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1311887-1312148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5483555 c.1766T>C p.L589P Chr.1: 1311914-1311914 on Build GRCh38
894854 c.1679C>T p.A560V Chr.1: 1312076-1312076 on Build GRCh38
6045092 c.1703C>T p.A568V Chr.1: 1312070-1312070 on Build GRCh38
7244371 c.1641A>G p.P547P Chr.1: 1312114-1312114 on Build GRCh38
6388201 c.1639C>A p.H547N Chr.1: 1312134-1312134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781305263 Chr.1: 1312129-1312129 on Build GRCh38
rs769879497 Chr.1: 1311948-1311948 on Build GRCh38
rs781337148 Chr.1: 1311984-1311984 on Build GRCh38
rs374968976 Chr.1: 1311964-1311964 on Build GRCh38
rs148516711 Chr.1: 1312111-1312111 on Build GRCh38
rs746897636 Chr.1: 1312116-1312116 on Build GRCh38
rs1003161341 Chr.1: 1312034-1312034 on Build GRCh38
rs201530038 Chr.1: 1311971-1311971 on Build GRCh38
rs71578312 Chr.1: 1311996-1311996 on Build GRCh38
rs755782365 Chr.1: 1311918-1311918 on Build GRCh38
rs201650418 Chr.1: 1312063-1312063 on Build GRCh38
rs751486747 Chr.1: 1312126-1312127 on Build GRCh38
rs893875851 Chr.1: 1311978-1311980 on Build GRCh38
rs146230519 Chr.1: 1312074-1312074 on Build GRCh38
rs767332000 Chr.1: 1311959-1311959 on Build GRCh38
rs374799704 Chr.1: 1312103-1312103 on Build GRCh38
rs767846766 Chr.1: 1312109-1312109 on Build GRCh38
rs750087288 Chr.1: 1311916-1311916 on Build GRCh38
rs757953455 Chr.1: 1312037-1312037 on Build GRCh38
rs772931868 Chr.1: 1311960-1311960 on Build GRCh38
rs771944123 Chr.1: 1311993-1311993 on Build GRCh38
rs758875060 Chr.1: 1311972-1311972 on Build GRCh38
rs200085739 Chr.1: 1311997-1311997 on Build GRCh38
rs200942028 Chr.1: 1311991-1311991 on Build GRCh38
rs374446194 Chr.1: 1311946-1311946 on Build GRCh38
rs200186070 Chr.1: 1311928-1311928 on Build GRCh38
rs12103 Chr.1: 1312114-1312114 on Build GRCh38
rs373702367 Chr.1: 1311929-1311929 on Build GRCh38
rs754469585 Chr.1: 1311969-1311969 on Build GRCh38
rs756503541 Chr.1: 1312110-1312110 on Build GRCh38
rs182929670 Chr.1: 1312090-1312090 on Build GRCh38
rs745607228 Chr.1: 1312131-1312131 on Build GRCh38
rs757501026 Chr.1: 1312054-1312054 on Build GRCh38
rs560124260 Chr.1: 1312108-1312108 on Build GRCh38
rs747230069 Chr.1: 1311936-1311936 on Build GRCh38
rs774210372 Chr.1: 1311950-1311950 on Build GRCh38
rs564287594 Chr.1: 1311949-1311949 on Build GRCh38
rs367622194 Chr.1: 1311987-1311987 on Build GRCh38
rs759319376 Chr.1: 1312012-1312012 on Build GRCh38
rs879100926 Chr.1: 1311979-1311980 on Build GRCh38
rs747032106 Chr.1: 1312075-1312075 on Build GRCh38
rs544280478 Chr.1: 1312035-1312035 on Build GRCh38
rs775845192 Chr.1: 1312085-1312086 on Build GRCh38
rs754557285 Chr.1: 1311924-1311924 on Build GRCh38
rs763140442 Chr.1: 1312094-1312095 on Build GRCh38
rs756378727 Chr.1: 1312064-1312064 on Build GRCh38
rs771094080 Chr.1: 1311944-1311944 on Build GRCh38
rs771029339 Chr.1: 1312126-1312126 on Build GRCh38
rs948085205 Chr.1: 1312122-1312122 on Build GRCh38
rs375229767 Chr.1: 1311992-1311992 on Build GRCh38
rs776865790 Chr.1: 1311945-1311945 on Build GRCh38
rs764207158 Chr.1: 1312096-1312097 on Build GRCh38
rs761808624 Chr.1: 1311955-1311955 on Build GRCh38
rs1027229991 Chr.1: 1312091-1312091 on Build GRCh38
rs1040629078 Chr.1: 1312002-1312002 on Build GRCh38
rs141621930 Chr.1: 1312039-1312039 on Build GRCh38
rs376179762 Chr.1: 1311935-1311935 on Build GRCh38
rs372283691 Chr.1: 1312031-1312031 on Build GRCh38
rs753557621 Chr.1: 1311965-1311965 on Build GRCh38
rs369652778 Chr.1: 1312099-1312099 on Build GRCh38
rs771218731 Chr.1: 1311962-1311962 on Build GRCh38
rs368386090 Chr.1: 1311994-1311994 on Build GRCh38
rs776417073 Chr.1: 1312000-1312000 on Build GRCh38
rs932999699 Chr.1: 1312098-1312098 on Build GRCh38
rs779503701 Chr.1: 1311925-1311925 on Build GRCh38
rs140077512 Chr.1: 1312089-1312089 on Build GRCh38
rs767224126 Chr.1: 1311908-1311908 on Build GRCh38
rs781606711 Chr.1: 1312058-1312058 on Build GRCh38
rs1042289398 Chr.1: 1312030-1312030 on Build GRCh38
rs759945647 Chr.1: 1312001-1312002 on Build GRCh38
rs199870169 Chr.1: 1311962-1311962 on Build GRCh38
rs370279028 Chr.1: 1311953-1311953 on Build GRCh38
rs375917361 Chr.1: 1312071-1312071 on Build GRCh38
rs770835204 Chr.1: 1312081-1312081 on Build GRCh38
rs377117800 Chr.1: 1311923-1311923 on Build GRCh38
rs764814488 Chr.1: 1312013-1312013 on Build GRCh38
rs908956003 Chr.1: 1311921-1311921 on Build GRCh38
rs765619076 Chr.1: 1312007-1312007 on Build GRCh38
rs777098288 Chr.1: 1311963-1311964 on Build GRCh38
rs752008859 Chr.1: 1312106-1312106 on Build GRCh38
rs776613388 Chr.1: 1312087-1312087 on Build GRCh38
rs923072220 Chr.1: 1312005-1312005 on Build GRCh38
rs778164963 Chr.1: 1311974-1311974 on Build GRCh38
rs563061550 Chr.1: 1311996-1311996 on Build GRCh38
rs146699248 Chr.1: 1311922-1311922 on Build GRCh38
rs755005813 Chr.1: 1312115-1312115 on Build GRCh38
rs775226496 Chr.1: 1312093-1312093 on Build GRCh38
rs752124216 Chr.1: 1312042-1312042 on Build GRCh38
rs945284202 Chr.1: 1312025-1312025 on Build GRCh38
rs138628609 Chr.1: 1312072-1312072 on Build GRCh38
rs747292136 Chr.1: 1311980-1311980 on Build GRCh38

View Full Product Details