Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 1057222-1057475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
UniGene
Hs.59506
Gene Chromosome Location
Chr.9: 1003237-1057554 on Build GRCh38
Transcripts
NM_001130865.2 NM_006557.6 NM_181872.4 XM_011517687.1 XM_011517690.2 XM_011517694.2 XM_017014213.1 XM_017014214.1 XM_017014215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779490 c.1636G>C p.E546Q Chr.9: 1057223-1057223 on Build GRCh38
1103410 c.1652G>T p.R551M Chr.9: 1057239-1057239 on Build GRCh38
5590564 c.1672C>T p.R558C Chr.9: 1057259-1057259 on Build GRCh38
1241084 c.1663G>A p.A555T Chr.9: 1057250-1057250 on Build GRCh38
5430887 c.1674T>A p.R558R Chr.9: 1057261-1057261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777304682 Chr.9: 1057322-1057322 on Build GRCh38
rs755526203 Chr.9: 1057252-1057252 on Build GRCh38
rs775343120 Chr.9: 1057287-1057287 on Build GRCh38
rs371831083 Chr.9: 1057250-1057250 on Build GRCh38
rs749806998 Chr.9: 1057249-1057249 on Build GRCh38
rs4741030 Chr.9: 1057351-1057351 on Build GRCh38
rs928010438 Chr.9: 1057270-1057270 on Build GRCh38
rs994917649 Chr.9: 1057399-1057399 on Build GRCh38
rs931888410 Chr.9: 1057335-1057335 on Build GRCh38
rs1003674743 Chr.9: 1057437-1057437 on Build GRCh38
rs772480908 Chr.9: 1057260-1057261 on Build GRCh38
rs765155889 Chr.9: 1057280-1057280 on Build GRCh38
rs149488494 Chr.9: 1057331-1057331 on Build GRCh38
rs778629637 Chr.9: 1057253-1057253 on Build GRCh38
rs771650790 Chr.9: 1057257-1057257 on Build GRCh38
rs760926881 Chr.9: 1057303-1057303 on Build GRCh38
rs543418900 Chr.9: 1057324-1057324 on Build GRCh38
rs773732202 Chr.9: 1057263-1057264 on Build GRCh38
rs776466458 Chr.9: 1057267-1057267 on Build GRCh38
rs902608185 Chr.9: 1057387-1057387 on Build GRCh38
rs770673047 Chr.9: 1057260-1057260 on Build GRCh38
rs761898433 Chr.9: 1057291-1057291 on Build GRCh38
rs759527693 Chr.9: 1057279-1057279 on Build GRCh38
rs779575171 Chr.9: 1057317-1057317 on Build GRCh38
rs41265270 Chr.9: 1057327-1057327 on Build GRCh38
rs764328228 Chr.9: 1057259-1057259 on Build GRCh38
rs371495993 Chr.9: 1057359-1057359 on Build GRCh38
rs368404730 Chr.9: 1057307-1057307 on Build GRCh38
rs563421536 Chr.9: 1057325-1057325 on Build GRCh38
rs202045027 Chr.9: 1057277-1057277 on Build GRCh38
rs754221398 Chr.9: 1057306-1057306 on Build GRCh38
rs1004569305 Chr.9: 1057361-1057361 on Build GRCh38
rs72703239 Chr.9: 1057294-1057294 on Build GRCh38
rs995944842 Chr.9: 1057342-1057342 on Build GRCh38
rs921810793 Chr.9: 1057335-1057336 on Build GRCh38
rs1015124645 Chr.9: 1057366-1057366 on Build GRCh38
rs747488837 Chr.9: 1057311-1057311 on Build GRCh38
rs9140 Chr.9: 1057318-1057318 on Build GRCh38
rs890313791 Chr.9: 1057360-1057360 on Build GRCh38
rs758011765 Chr.9: 1057321-1057321 on Build GRCh38
rs374340988 Chr.9: 1057297-1057297 on Build GRCh38
rs963067437 Chr.9: 1057369-1057369 on Build GRCh38
rs1023849312 Chr.9: 1057424-1057424 on Build GRCh38
rs556197778 Chr.9: 1057256-1057256 on Build GRCh38
rs766709953 Chr.9: 1057305-1057305 on Build GRCh38

View Full Product Details