Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27368507-27368770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983035 c.505C>G p.Q169E Chr.2: 27368707-27368707 on Build GRCh38
5955656 c.490C>T p.R164C Chr.2: 27368662-27368662 on Build GRCh38
5955655 c.550C>T p.R184C Chr.2: 27368662-27368662 on Build GRCh38
4983036 c.445C>G p.Q149E Chr.2: 27368707-27368707 on Build GRCh38
211973 c.451G>C p.D151H Chr.2: 27368701-27368701 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201503266 Chr.2: 27368666-27368666 on Build GRCh38
rs569664227 Chr.2: 27368697-27368697 on Build GRCh38
rs913265804 Chr.2: 27368538-27368538 on Build GRCh38
rs1008673121 Chr.2: 27368647-27368647 on Build GRCh38
rs752260211 Chr.2: 27368745-27368745 on Build GRCh38
rs777786550 Chr.2: 27368644-27368644 on Build GRCh38
rs201042063 Chr.2: 27368640-27368640 on Build GRCh38
rs754559917 Chr.2: 27368599-27368599 on Build GRCh38
rs758627794 Chr.2: 27368582-27368582 on Build GRCh38
rs750850634 Chr.2: 27368719-27368719 on Build GRCh38
rs886038382 Chr.2: 27368617-27368617 on Build GRCh38
rs947539380 Chr.2: 27368570-27368570 on Build GRCh38
rs866357253 Chr.2: 27368670-27368670 on Build GRCh38
rs751135430 Chr.2: 27368588-27368588 on Build GRCh38
rs758641876 Chr.2: 27368535-27368535 on Build GRCh38
rs779565150 Chr.2: 27368588-27368589 on Build GRCh38
rs570777761 Chr.2: 27368740-27368740 on Build GRCh38
rs761810644 Chr.2: 27368711-27368711 on Build GRCh38
rs370413772 Chr.2: 27368650-27368650 on Build GRCh38
rs770591078 Chr.2: 27368661-27368661 on Build GRCh38
rs914750494 Chr.2: 27368562-27368562 on Build GRCh38
rs775819795 Chr.2: 27368662-27368662 on Build GRCh38
rs779235094 Chr.2: 27368744-27368746 on Build GRCh38
rs529735415 Chr.2: 27368639-27368639 on Build GRCh38
rs765743112 Chr.2: 27368714-27368714 on Build GRCh38
rs147672203 Chr.2: 27368669-27368669 on Build GRCh38
rs149121402 Chr.2: 27368723-27368723 on Build GRCh38
rs377518092 Chr.2: 27368620-27368620 on Build GRCh38
rs754665816 Chr.2: 27368725-27368725 on Build GRCh38
rs946111331 Chr.2: 27368552-27368552 on Build GRCh38
rs779782574 Chr.2: 27368583-27368583 on Build GRCh38
rs748155307 Chr.2: 27368634-27368634 on Build GRCh38
rs368243788 Chr.2: 27368724-27368724 on Build GRCh38
rs750788095 Chr.2: 27368579-27368579 on Build GRCh38
rs768779136 Chr.2: 27368685-27368685 on Build GRCh38
rs776685186 Chr.2: 27368695-27368695 on Build GRCh38
rs1053334796 Chr.2: 27368540-27368540 on Build GRCh38

View Full Product Details