Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1326754-1326981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPTP
Entrez Gene ID
80772
Gene Name
ceramide-1-phosphate transfer protein
Gene Aliases
GLTPD1
UniGene
Hs.515689
Gene Chromosome Location
Chr.1: 1324756-1328896 on Build GRCh38
Transcripts
NM_001029885.1 XM_005244801.3 XM_005244802.1 XM_011542200.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333360 c.12G>A p.S4S Chr.1: 1326922-1326922 on Build GRCh38
6671845 c.63C>T p.L21L Chr.1: 1326973-1326973 on Build GRCh38
3740593 c.1A>G p.M1V Chr.1: 1326911-1326911 on Build GRCh38
4687283 c.25A>G p.N9D Chr.1: 1326935-1326935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747107838 Chr.1: 1326879-1326879 on Build GRCh38
rs368856116 Chr.1: 1326867-1326867 on Build GRCh38
rs1028726129 Chr.1: 1326775-1326775 on Build GRCh38
rs537592194 Chr.1: 1326796-1326796 on Build GRCh38
rs747947774 Chr.1: 1326897-1326897 on Build GRCh38
rs956644988 Chr.1: 1326823-1326823 on Build GRCh38
rs904112602 Chr.1: 1326794-1326794 on Build GRCh38
rs568610024 Chr.1: 1326784-1326784 on Build GRCh38
rs1001160690 Chr.1: 1326806-1326806 on Build GRCh38
rs908696029 Chr.1: 1326901-1326901 on Build GRCh38
rs201512938 Chr.1: 1326955-1326955 on Build GRCh38
rs757321048 Chr.1: 1326881-1326881 on Build GRCh38
rs371310951 Chr.1: 1326922-1326922 on Build GRCh38
rs571561984 Chr.1: 1326811-1326811 on Build GRCh38
rs901484111 Chr.1: 1326774-1326774 on Build GRCh38
rs973146164 Chr.1: 1326891-1326891 on Build GRCh38
rs928047686 Chr.1: 1326883-1326883 on Build GRCh38
rs754390803 Chr.1: 1326882-1326882 on Build GRCh38
rs576563684 Chr.1: 1326921-1326921 on Build GRCh38
rs751777867 Chr.1: 1326947-1326947 on Build GRCh38
rs554104587 Chr.1: 1326837-1326837 on Build GRCh38
rs185479627 Chr.1: 1326919-1326919 on Build GRCh38
rs772756143 Chr.1: 1326936-1326936 on Build GRCh38
rs549990835 Chr.1: 1326778-1326778 on Build GRCh38
rs374389586 Chr.1: 1326877-1326877 on Build GRCh38
rs763295581 Chr.1: 1326930-1326930 on Build GRCh38
rs113667133 Chr.1: 1326896-1326896 on Build GRCh38
rs760341483 Chr.1: 1326940-1326940 on Build GRCh38
rs143006705 Chr.1: 1326874-1326874 on Build GRCh38
rs766011321 Chr.1: 1326951-1326951 on Build GRCh38
rs755486100 Chr.1: 1326903-1326904 on Build GRCh38
rs538937809 Chr.1: 1326884-1326884 on Build GRCh38
rs193084995 Chr.1: 1326862-1326862 on Build GRCh38
rs781213133 Chr.1: 1326949-1326949 on Build GRCh38
rs760196521 Chr.1: 1326907-1326907 on Build GRCh38
rs930546989 Chr.1: 1326913-1326913 on Build GRCh38
rs771740123 Chr.1: 1326902-1326902 on Build GRCh38
rs764346804 Chr.1: 1326946-1326946 on Build GRCh38
rs778877504 Chr.1: 1326953-1326953 on Build GRCh38
rs1008704892 Chr.1: 1326782-1326782 on Build GRCh38
rs534068993 Chr.1: 1326866-1326866 on Build GRCh38
rs377591659 Chr.1: 1326886-1326886 on Build GRCh38
rs754854819 Chr.1: 1326952-1326952 on Build GRCh38
rs772803583 Chr.1: 1326905-1326905 on Build GRCh38
rs138651562 Chr.1: 1326882-1326882 on Build GRCh38
rs373239114 Chr.1: 1326912-1326912 on Build GRCh38

View Full Product Details