Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102259909-102260152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBF1
Entrez Gene ID
8729
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
Gene Aliases
ARF1GEF
UniGene
Hs.290243
Gene Chromosome Location
Chr.10: 102245498-102382899 on Build GRCh38
Transcripts
NM_001199378.1 NM_001199379.1 NM_004193.2 XM_005270261.1 XM_006718047.1 XM_006718048.1 XM_006718049.1 XM_006718050.1 XM_011540312.1 XM_011540313.1 XM_017016861.1 XM_017016862.1 XM_017016863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914148 c.106C>T p.R36W Chr.10: 102260059-102260059 on Build GRCh38
3978044 c.155G>C p.S52T Chr.10: 102260108-102260108 on Build GRCh38
6194157 c.144G>A p.E48E Chr.10: 102260097-102260097 on Build GRCh38
7198809 c.125G>A p.S42N Chr.10: 102260078-102260078 on Build GRCh38
1504200 c.136C>G p.L46V Chr.10: 102260089-102260089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756702754 Chr.10: 102260059-102260059 on Build GRCh38
rs74155240 Chr.10: 102259941-102259941 on Build GRCh38
rs201533615 Chr.10: 102260060-102260060 on Build GRCh38
rs766681048 Chr.10: 102260019-102260019 on Build GRCh38
rs761209886 Chr.10: 102260017-102260017 on Build GRCh38
rs1032218849 Chr.10: 102260004-102260004 on Build GRCh38
rs752377654 Chr.10: 102260121-102260123 on Build GRCh38
rs34665033 Chr.10: 102260068-102260069 on Build GRCh38
rs868229634 Chr.10: 102260093-102260093 on Build GRCh38
rs761113491 Chr.10: 102260113-102260113 on Build GRCh38
rs116242655 Chr.10: 102259999-102259999 on Build GRCh38
rs544769042 Chr.10: 102260114-102260114 on Build GRCh38
rs756138137 Chr.10: 102260066-102260066 on Build GRCh38
rs913643879 Chr.10: 102260020-102260020 on Build GRCh38
rs754216437 Chr.10: 102260026-102260026 on Build GRCh38
rs925208130 Chr.10: 102259997-102259997 on Build GRCh38
rs192723377 Chr.10: 102259959-102259959 on Build GRCh38
rs777289121 Chr.10: 102260051-102260051 on Build GRCh38
rs758004436 Chr.10: 102260042-102260042 on Build GRCh38
rs773311713 Chr.10: 102260110-102260110 on Build GRCh38
rs577447540 Chr.10: 102260082-102260082 on Build GRCh38
rs772107449 Chr.10: 102260104-102260104 on Build GRCh38
rs768560722 Chr.10: 102260083-102260083 on Build GRCh38
rs148913915 Chr.10: 102259947-102259947 on Build GRCh38
rs553027274 Chr.10: 102260065-102260065 on Build GRCh38
rs779933281 Chr.10: 102260075-102260075 on Build GRCh38
rs751286161 Chr.10: 102260056-102260056 on Build GRCh38
rs765045197 Chr.10: 102260019-102260019 on Build GRCh38
rs748450100 Chr.10: 102260091-102260091 on Build GRCh38

View Full Product Details