Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1984870-1985099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFER
Entrez Gene ID
2671
Gene Name
growth factor, augmenter of liver regeneration
Gene Aliases
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
UniGene
Hs.27184
Gene Chromosome Location
Chr.16: 1984149-1987749 on Build GRCh38
Transcripts
NM_005262.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895365 c.427G>C p.E143Q Chr.16: 1984915-1984915 on Build GRCh38
1678758 c.410C>A p.S137Y Chr.16: 1984898-1984898 on Build GRCh38
6362566 c.422C>T p.P141L Chr.16: 1984910-1984910 on Build GRCh38
6360239 c.395T>C p.F132S Chr.16: 1984883-1984883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991504262 Chr.16: 1985043-1985043 on Build GRCh38
rs929832047 Chr.16: 1985068-1985068 on Build GRCh38
rs750605745 Chr.16: 1984988-1984988 on Build GRCh38
rs376478434 Chr.16: 1984995-1984995 on Build GRCh38
rs750515991 Chr.16: 1984917-1984917 on Build GRCh38
rs863224026 Chr.16: 1984942-1984942 on Build GRCh38
rs756436660 Chr.16: 1984991-1984991 on Build GRCh38
rs369886365 Chr.16: 1984933-1984933 on Build GRCh38
rs760336801 Chr.16: 1984964-1984964 on Build GRCh38
rs1046764757 Chr.16: 1985069-1985069 on Build GRCh38
rs770384334 Chr.16: 1984970-1984970 on Build GRCh38
rs757388602 Chr.16: 1984911-1984911 on Build GRCh38
rs373446345 Chr.16: 1984910-1984910 on Build GRCh38
rs571384984 Chr.16: 1985046-1985046 on Build GRCh38
rs898325421 Chr.16: 1985065-1985065 on Build GRCh38
rs763341760 Chr.16: 1984978-1984978 on Build GRCh38
rs928741231 Chr.16: 1985015-1985015 on Build GRCh38
rs781628694 Chr.16: 1984913-1984913 on Build GRCh38
rs762295209 Chr.16: 1984986-1984986 on Build GRCh38
rs534308773 Chr.16: 1985056-1985056 on Build GRCh38
rs756203483 Chr.16: 1984925-1984925 on Build GRCh38
rs764697486 Chr.16: 1984980-1984980 on Build GRCh38
rs748018775 Chr.16: 1984955-1984955 on Build GRCh38
rs538091471 Chr.16: 1984976-1984976 on Build GRCh38
rs569048447 Chr.16: 1984952-1984952 on Build GRCh38
rs923068897 Chr.16: 1985015-1985016 on Build GRCh38
rs768661163 Chr.16: 1984951-1984951 on Build GRCh38
rs3177436 Chr.16: 1984945-1984945 on Build GRCh38
rs771712671 Chr.16: 1984960-1984960 on Build GRCh38
rs931891068 Chr.16: 1985022-1985022 on Build GRCh38
rs778160972 Chr.16: 1984961-1984961 on Build GRCh38
rs1038714161 Chr.16: 1985058-1985058 on Build GRCh38
rs916469728 Chr.16: 1985045-1985045 on Build GRCh38
rs938632533 Chr.16: 1985016-1985016 on Build GRCh38
rs780287282 Chr.16: 1984946-1984946 on Build GRCh38
rs763545328 Chr.16: 1984909-1984909 on Build GRCh38
rs549270205 Chr.16: 1984949-1984949 on Build GRCh38

View Full Product Details