Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206771703-206771952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
UniGene
Hs.5930
Gene Chromosome Location
Chr.2: 206765388-206868284 on Build GRCh38
Transcripts
NM_001136193.1 NM_001136194.1 NM_014929.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291421 c.1004G>T p.R335M Chr.2: 206771907-206771907 on Build GRCh38
5531043 c.1022C>T p.S341F Chr.2: 206771925-206771925 on Build GRCh38
3909419 c.999C>T p.A333A Chr.2: 206771902-206771902 on Build GRCh38
5611718 c.991-2A>T p.? Chr.2: 206771892-206771892 on Build GRCh38
6660703 c.1018T>G p.F340V Chr.2: 206771921-206771921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774891673 Chr.2: 206771892-206771892 on Build GRCh38
rs1036908790 Chr.2: 206771783-206771783 on Build GRCh38
rs568793725 Chr.2: 206771784-206771784 on Build GRCh38
rs566208529 Chr.2: 206771739-206771739 on Build GRCh38
rs774329789 Chr.2: 206771919-206771919 on Build GRCh38
rs762491175 Chr.2: 206771911-206771911 on Build GRCh38
rs772556733 Chr.2: 206771855-206771855 on Build GRCh38
rs915111997 Chr.2: 206771798-206771798 on Build GRCh38
rs1005461541 Chr.2: 206771751-206771751 on Build GRCh38
rs187368105 Chr.2: 206771776-206771776 on Build GRCh38
rs989224953 Chr.2: 206771796-206771796 on Build GRCh38
rs769518431 Chr.2: 206771891-206771891 on Build GRCh38
rs763986729 Chr.2: 206771917-206771917 on Build GRCh38
rs775962617 Chr.2: 206771904-206771905 on Build GRCh38
rs532206253 Chr.2: 206771750-206771750 on Build GRCh38
rs13392746 Chr.2: 206771826-206771826 on Build GRCh38
rs759893830 Chr.2: 206771848-206771881 on Build GRCh38
rs927842754 Chr.2: 206771844-206771844 on Build GRCh38
rs13421046 Chr.2: 206771881-206771881 on Build GRCh38
rs772300042 Chr.2: 206771900-206771902 on Build GRCh38
rs554844533 Chr.2: 206771806-206771806 on Build GRCh38
rs751398649 Chr.2: 206771822-206771822 on Build GRCh38
rs12620432 Chr.2: 206771794-206771794 on Build GRCh38
rs1019576676 Chr.2: 206771745-206771745 on Build GRCh38
rs965093188 Chr.2: 206771746-206771746 on Build GRCh38

View Full Product Details