Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179043829-179044092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20B
Entrez Gene ID
9917
Gene Name
family with sequence similarity 20 member B
Gene Aliases
gxk1
UniGene
Hs.5737
Gene Chromosome Location
Chr.1: 179025804-179076574 on Build GRCh38
Transcripts
NM_001324310.1 NM_001324311.1 NM_014864.3 XM_017003001.1 XM_017003002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900189 c.44T>C p.V15A Chr.1: 179043891-179043891 on Build GRCh38
424689 c.21C>T p.V7V Chr.1: 179043868-179043868 on Build GRCh38
5696796 c.27G>C p.L9L Chr.1: 179043874-179043874 on Build GRCh38
6873783 c.207G>A p.Q69Q Chr.1: 179044054-179044054 on Build GRCh38
5901030 c.73A>T p.I25F Chr.1: 179043920-179043920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754757784 Chr.1: 179044023-179044023 on Build GRCh38
rs754880630 Chr.1: 179043926-179043926 on Build GRCh38
rs375526878 Chr.1: 179044034-179044034 on Build GRCh38
rs763729864 Chr.1: 179043878-179043878 on Build GRCh38
rs1024965514 Chr.1: 179044005-179044005 on Build GRCh38
rs771439887 Chr.1: 179043963-179043963 on Build GRCh38
rs971326119 Chr.1: 179044031-179044031 on Build GRCh38
rs561266417 Chr.1: 179043976-179043976 on Build GRCh38
rs777032848 Chr.1: 179043868-179043868 on Build GRCh38
rs1008960850 Chr.1: 179044020-179044020 on Build GRCh38
rs749107151 Chr.1: 179043924-179043924 on Build GRCh38
rs1009034510 Chr.1: 179043996-179043996 on Build GRCh38
rs146067221 Chr.1: 179043951-179043951 on Build GRCh38
rs895722601 Chr.1: 179043869-179043869 on Build GRCh38
rs781681473 Chr.1: 179044070-179044070 on Build GRCh38
rs749923727 Chr.1: 179043992-179043992 on Build GRCh38
rs766961096 Chr.1: 179043979-179043979 on Build GRCh38
rs200886195 Chr.1: 179043966-179043966 on Build GRCh38
rs545932932 Chr.1: 179043907-179043907 on Build GRCh38
rs746216042 Chr.1: 179044068-179044068 on Build GRCh38
rs756697625 Chr.1: 179043893-179043893 on Build GRCh38
rs777898569 Chr.1: 179044055-179044055 on Build GRCh38
rs773926718 Chr.1: 179043974-179043974 on Build GRCh38
rs759930487 Chr.1: 179043965-179043965 on Build GRCh38
rs779222538 Chr.1: 179043927-179043927 on Build GRCh38
rs940560627 Chr.1: 179043917-179043917 on Build GRCh38
rs1002787984 Chr.1: 179044035-179044035 on Build GRCh38
rs766330375 Chr.1: 179044001-179044001 on Build GRCh38
rs768467670 Chr.1: 179043911-179043911 on Build GRCh38
rs755617315 Chr.1: 179043993-179043993 on Build GRCh38
rs892527665 Chr.1: 179043950-179043950 on Build GRCh38
rs759856276 Chr.1: 179043875-179043875 on Build GRCh38
rs751271045 Chr.1: 179043881-179043881 on Build GRCh38
rs370513530 Chr.1: 179043928-179043928 on Build GRCh38
rs773449058 Chr.1: 179043948-179043948 on Build GRCh38
rs137869702 Chr.1: 179043923-179043923 on Build GRCh38
rs766605797 Chr.1: 179043864-179043864 on Build GRCh38
rs760272283 Chr.1: 179043884-179043886 on Build GRCh38
rs370939998 Chr.1: 179043856-179043856 on Build GRCh38
rs753735977 Chr.1: 179044008-179044008 on Build GRCh38
rs574739267 Chr.1: 179044044-179044044 on Build GRCh38
rs772239388 Chr.1: 179043934-179043934 on Build GRCh38
rs11802142 Chr.1: 179044069-179044069 on Build GRCh38
rs142026121 Chr.1: 179043975-179043975 on Build GRCh38
rs767015961 Chr.1: 179043905-179043905 on Build GRCh38
rs778883078 Chr.1: 179044032-179044032 on Build GRCh38

View Full Product Details