Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129067722-129067953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIGA2
Entrez Gene ID
84895
Gene Name
mitoguardin 2
Gene Aliases
C9orf54, FAM73B
UniGene
Hs.632693
Gene Chromosome Location
Chr.9: 129036605-129072082 on Build GRCh38
Transcripts
NM_032809.2 XM_005252282.1 XM_011519117.1 XM_011519118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654786 c.1264C>T p.R422* Chr.9: 129067866-129067866 on Build GRCh38
6550510 c.1203G>T p.E401D Chr.9: 129067805-129067805 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs147360024 Chr.9: 129067757-129067757 on Build GRCh38
rs764836417 Chr.9: 129067784-129067784 on Build GRCh38
rs750157229 Chr.9: 129067827-129067827 on Build GRCh38
rs763746025 Chr.9: 129067896-129067896 on Build GRCh38
rs769623185 Chr.9: 129067856-129067856 on Build GRCh38
rs767338421 Chr.9: 129067909-129067909 on Build GRCh38
rs767526015 Chr.9: 129067824-129067824 on Build GRCh38
rs761132275 Chr.9: 129067780-129067780 on Build GRCh38
rs761429099 Chr.9: 129067904-129067904 on Build GRCh38
rs369949431 Chr.9: 129067807-129067807 on Build GRCh38
rs753699705 Chr.9: 129067920-129067920 on Build GRCh38
rs756024642 Chr.9: 129067829-129067829 on Build GRCh38
rs771340663 Chr.9: 129067891-129067891 on Build GRCh38
rs550220083 Chr.9: 129067819-129067819 on Build GRCh38
rs773807904 Chr.9: 129067870-129067870 on Build GRCh38
rs755971278 Chr.9: 129067910-129067910 on Build GRCh38
rs1018154753 Chr.9: 129067814-129067814 on Build GRCh38
rs749345752 Chr.9: 129067753-129067753 on Build GRCh38
rs200646327 Chr.9: 129067802-129067802 on Build GRCh38
rs570344208 Chr.9: 129067895-129067895 on Build GRCh38
rs935025600 Chr.9: 129067867-129067867 on Build GRCh38
rs779683609 Chr.9: 129067900-129067900 on Build GRCh38
rs747350331 Chr.9: 129067888-129067888 on Build GRCh38
rs752394279 Chr.9: 129067799-129067799 on Build GRCh38
rs998271905 Chr.9: 129067853-129067853 on Build GRCh38
rs755257337 Chr.9: 129067859-129067859 on Build GRCh38
rs775115715 Chr.9: 129067894-129067894 on Build GRCh38
rs772417964 Chr.9: 129067866-129067866 on Build GRCh38
rs979185093 Chr.9: 129067818-129067818 on Build GRCh38
rs778800545 Chr.9: 129067861-129067861 on Build GRCh38
rs780412145 Chr.9: 129067846-129067846 on Build GRCh38
rs149779596 Chr.9: 129067787-129067787 on Build GRCh38
rs749590870 Chr.9: 129067854-129067854 on Build GRCh38
rs768921850 Chr.9: 129067756-129067756 on Build GRCh38
rs766211390 Chr.9: 129067918-129067918 on Build GRCh38
rs774037538 Chr.9: 129067792-129067792 on Build GRCh38
rs773400349 Chr.9: 129067769-129067769 on Build GRCh38
rs748282219 Chr.9: 129067863-129067863 on Build GRCh38
rs751081518 Chr.9: 129067817-129067817 on Build GRCh38
rs1006798871 Chr.9: 129067762-129067762 on Build GRCh38
rs200120093 Chr.9: 129067931-129067931 on Build GRCh38

View Full Product Details