Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141516914-141517139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH1
Entrez Gene ID
1729
Gene Name
diaphanous related formin 1
Gene Aliases
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
UniGene
Hs.529451
Gene Chromosome Location
Chr.5: 141515021-141619055 on Build GRCh38
Transcripts
NM_001079812.2 NM_001314007.1 NM_005219.4 XM_011537572.2 XM_011537573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5788638 c.3662-4G>C p.? Chr.5: 141517012-141517012 on Build GRCh38
3852213 c.3682G>A p.A1228T Chr.5: 141516988-141516988 on Build GRCh38
5680841 c.3696G>T p.L1232L Chr.5: 141516974-141516974 on Build GRCh38
1496017 c.3663C>A p.A1221A Chr.5: 141517007-141517007 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs925999346 Chr.5: 141517018-141517018 on Build GRCh38
rs541845126 Chr.5: 141517023-141517023 on Build GRCh38
rs371385202 Chr.5: 141516995-141516995 on Build GRCh38
rs371640300 Chr.5: 141517025-141517025 on Build GRCh38
rs560824183 Chr.5: 141517024-141517024 on Build GRCh38
rs774815702 Chr.5: 141516939-141516939 on Build GRCh38
rs376143083 Chr.5: 141517040-141517040 on Build GRCh38
rs752983682 Chr.5: 141517011-141517011 on Build GRCh38
rs749871871 Chr.5: 141516934-141516934 on Build GRCh38
rs376832359 Chr.5: 141516999-141516999 on Build GRCh38
rs778069558 Chr.5: 141517015-141517015 on Build GRCh38
rs749920378 Chr.5: 141516978-141516978 on Build GRCh38
rs747855273 Chr.5: 141517057-141517057 on Build GRCh38
rs369935242 Chr.5: 141517022-141517022 on Build GRCh38
rs764739820 Chr.5: 141516969-141516969 on Build GRCh38
rs777476610 Chr.5: 141517030-141517033 on Build GRCh38
rs368448851 Chr.5: 141517029-141517029 on Build GRCh38
rs775772109 Chr.5: 141517038-141517038 on Build GRCh38
rs759795988 Chr.5: 141516947-141516947 on Build GRCh38
rs761387295 Chr.5: 141516966-141516966 on Build GRCh38
rs72790081 Chr.5: 141517032-141517032 on Build GRCh38
rs779307746 Chr.5: 141517026-141517026 on Build GRCh38
rs756395607 Chr.5: 141517012-141517012 on Build GRCh38
rs758159839 Chr.5: 141517023-141517023 on Build GRCh38
rs771335028 Chr.5: 141516935-141516935 on Build GRCh38
rs547600019 Chr.5: 141517087-141517087 on Build GRCh38
rs1008936590 Chr.5: 141517076-141517076 on Build GRCh38
rs772429379 Chr.5: 141516964-141516964 on Build GRCh38
rs908230316 Chr.5: 141517086-141517086 on Build GRCh38
rs776296177 Chr.5: 141516965-141516965 on Build GRCh38
rs772326558 Chr.5: 141517037-141517037 on Build GRCh38
rs766204786 Chr.5: 141516984-141516984 on Build GRCh38
rs754761532 Chr.5: 141516997-141516997 on Build GRCh38

View Full Product Details