Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76832656-76832889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL1
Entrez Gene ID
2150
Gene Name
F2R like trypsin receptor 1
Gene Aliases
GPR11, PAR2
UniGene
Hs.744181
Gene Chromosome Location
Chr.5: 76819008-76835315 on Build GRCh38
Transcripts
XM_017009223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3617956 c.194C>T p.S65F Chr.5: 76832801-76832801 on Build GRCh38
7317118 c.187G>A p.E63K Chr.5: 76832794-76832794 on Build GRCh38
3617957 c.231C>T p.F77F Chr.5: 76832838-76832838 on Build GRCh38
1196833 c.280G>T p.G94C Chr.5: 76832887-76832887 on Build GRCh38
4159992 c.89G>A p.S30N Chr.5: 76832696-76832696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs616235 Chr.5: 76832696-76832696 on Build GRCh38
rs775613107 Chr.5: 76832707-76832707 on Build GRCh38
rs1007877867 Chr.5: 76832749-76832749 on Build GRCh38
rs746271767 Chr.5: 76832828-76832828 on Build GRCh38
rs879102015 Chr.5: 76832771-76832771 on Build GRCh38
rs148375929 Chr.5: 76832748-76832748 on Build GRCh38
rs922971746 Chr.5: 76832829-76832829 on Build GRCh38
rs776777310 Chr.5: 76832697-76832697 on Build GRCh38
rs781567981 Chr.5: 76832827-76832827 on Build GRCh38
rs767479233 Chr.5: 76832710-76832710 on Build GRCh38
rs34308580 Chr.5: 76832824-76832824 on Build GRCh38
rs755903838 Chr.5: 76832757-76832757 on Build GRCh38
rs1040032970 Chr.5: 76832777-76832777 on Build GRCh38
rs149644507 Chr.5: 76832703-76832703 on Build GRCh38
rs754704702 Chr.5: 76832796-76832796 on Build GRCh38
rs770039524 Chr.5: 76832832-76832832 on Build GRCh38
rs2243061 Chr.5: 76832686-76832686 on Build GRCh38
rs141650701 Chr.5: 76832761-76832761 on Build GRCh38
rs115250856 Chr.5: 76832784-76832784 on Build GRCh38
rs769256567 Chr.5: 76832845-76832845 on Build GRCh38
rs773639707 Chr.5: 76832688-76832688 on Build GRCh38
rs752228507 Chr.5: 76832808-76832808 on Build GRCh38
rs769871661 Chr.5: 76832859-76832860 on Build GRCh38
rs768626598 Chr.5: 76832866-76832866 on Build GRCh38
rs147135859 Chr.5: 76832812-76832812 on Build GRCh38
rs761940932 Chr.5: 76832776-76832777 on Build GRCh38
rs199908265 Chr.5: 76832837-76832837 on Build GRCh38
rs747358596 Chr.5: 76832695-76832695 on Build GRCh38
rs775636310 Chr.5: 76832844-76832844 on Build GRCh38
rs997300892 Chr.5: 76832868-76832868 on Build GRCh38
rs759776923 Chr.5: 76832701-76832701 on Build GRCh38
rs762995620 Chr.5: 76832709-76832709 on Build GRCh38
rs757902001 Chr.5: 76832809-76832809 on Build GRCh38
rs143373085 Chr.5: 76832726-76832726 on Build GRCh38
rs115873467 Chr.5: 76832867-76832867 on Build GRCh38
rs866400242 Chr.5: 76832711-76832711 on Build GRCh38
rs766287060 Chr.5: 76832759-76832759 on Build GRCh38

View Full Product Details