Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42248969-42249208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERF
Entrez Gene ID
2077
Gene Name
ETS2 repressor factor
Gene Aliases
CRS4, PE-2, PE2
UniGene
Hs.655969
Gene Chromosome Location
Chr.19: 42247561-42255164 on Build GRCh38
Transcripts
NM_001301035.1 NM_001308402.1 NM_001312656.1 NM_006494.3 XM_017026468.1 XM_017026469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534665 c.1103C>T p.S368F Chr.19: 42249009-42249009 on Build GRCh38
6557951 c.1087T>A p.S363T Chr.19: 42249025-42249025 on Build GRCh38
1612324 c.909C>A p.F303L Chr.19: 42249203-42249203 on Build GRCh38
6656054 c.1119C>T p.S373S Chr.19: 42248993-42248993 on Build GRCh38
3534666 c.1085C>T p.S362F Chr.19: 42249027-42249027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923175877 Chr.19: 42248999-42248999 on Build GRCh38
rs755405849 Chr.19: 42249104-42249104 on Build GRCh38
rs768373176 Chr.19: 42249143-42249143 on Build GRCh38
rs76105803 Chr.19: 42249008-42249008 on Build GRCh38
rs746938827 Chr.19: 42249141-42249141 on Build GRCh38
rs752492976 Chr.19: 42249011-42249011 on Build GRCh38
rs61735151 Chr.19: 42249131-42249131 on Build GRCh38
rs772094252 Chr.19: 42249062-42249062 on Build GRCh38
rs776460587 Chr.19: 42249152-42249152 on Build GRCh38
rs747311260 Chr.19: 42249037-42249037 on Build GRCh38
rs763081786 Chr.19: 42249170-42249170 on Build GRCh38
rs759570663 Chr.19: 42249061-42249061 on Build GRCh38
rs780415692 Chr.19: 42249036-42249036 on Build GRCh38
rs761960986 Chr.19: 42249154-42249154 on Build GRCh38
rs377467874 Chr.19: 42249081-42249081 on Build GRCh38
rs538236832 Chr.19: 42249024-42249024 on Build GRCh38
rs763637628 Chr.19: 42248995-42248998 on Build GRCh38
rs752052933 Chr.19: 42249103-42249103 on Build GRCh38
rs761007034 Chr.19: 42249014-42249014 on Build GRCh38
rs577264310 Chr.19: 42249017-42249017 on Build GRCh38
rs199836815 Chr.19: 42249032-42249032 on Build GRCh38
rs781590022 Chr.19: 42249114-42249114 on Build GRCh38
rs773296866 Chr.19: 42249169-42249169 on Build GRCh38
rs372067864 Chr.19: 42249023-42249023 on Build GRCh38
rs370719643 Chr.19: 42249064-42249064 on Build GRCh38
rs913822192 Chr.19: 42249029-42249029 on Build GRCh38
rs756887571 Chr.19: 42249124-42249124 on Build GRCh38
rs758877628 Chr.19: 42249035-42249035 on Build GRCh38
rs755455023 Chr.19: 42249178-42249178 on Build GRCh38
rs754070619 Chr.19: 42249074-42249074 on Build GRCh38
rs762060326 Chr.19: 42249075-42249075 on Build GRCh38
rs757176564 Chr.19: 42248997-42249002 on Build GRCh38
rs144812092 Chr.19: 42249066-42249066 on Build GRCh38
rs767470579 Chr.19: 42249000-42249000 on Build GRCh38
rs780483224 Chr.19: 42249092-42249092 on Build GRCh38
rs778319851 Chr.19: 42249125-42249125 on Build GRCh38
rs768011203 Chr.19: 42249044-42249044 on Build GRCh38
rs752101971 Chr.19: 42249177-42249177 on Build GRCh38
rs748943380 Chr.19: 42249122-42249122 on Build GRCh38
rs745308883 Chr.19: 42249128-42249128 on Build GRCh38
rs1009543151 Chr.19: 42248996-42248997 on Build GRCh38
rs766445497 Chr.19: 42249171-42249171 on Build GRCh38
rs756407997 Chr.19: 42249082-42249087 on Build GRCh38
rs149405304 Chr.19: 42249055-42249055 on Build GRCh38
rs769453776 Chr.19: 42249047-42249047 on Build GRCh38
rs745600955 Chr.19: 42249009-42249011 on Build GRCh38
rs1049744926 Chr.19: 42249006-42249006 on Build GRCh38
rs763967813 Chr.19: 42249071-42249071 on Build GRCh38
rs372854370 Chr.19: 42249049-42249049 on Build GRCh38
rs370634155 Chr.19: 42249161-42249161 on Build GRCh38
rs753965187 Chr.19: 42249018-42249018 on Build GRCh38
rs186795992 Chr.19: 42249059-42249059 on Build GRCh38
rs897260918 Chr.19: 42249046-42249046 on Build GRCh38
rs758914076 Chr.19: 42249091-42249091 on Build GRCh38
rs750465267 Chr.19: 42249086-42249086 on Build GRCh38
rs199960550 Chr.19: 42248997-42248999 on Build GRCh38
rs376130330 Chr.19: 42249060-42249060 on Build GRCh38
rs781154235 Chr.19: 42249008-42249009 on Build GRCh38

View Full Product Details