Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44858995-44859266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFTUD2
Entrez Gene ID
9343
Gene Name
elongation factor Tu GTP binding domain containing 2
Gene Aliases
MFDGA, MFDM, SNRNP116, Snrp116, Snu114, U5-116KD
UniGene
Hs.151787
Gene Chromosome Location
Chr.17: 44850287-44899625 on Build GRCh38
Transcripts
NM_001142605.1 NM_001258353.1 NM_001258354.1 NM_004247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5852674 c.1934G>A p.R645Q Chr.17: 44859108-44859108 on Build GRCh38
5961610 c.1896T>G p.T632T Chr.17: 44859146-44859146 on Build GRCh38
1302901 c.1864G>C p.E622Q Chr.17: 44859178-44859178 on Build GRCh38
5055518 c.1933C>T p.R645W Chr.17: 44859109-44859109 on Build GRCh38
6322169 c.1861-3C>A p.? Chr.17: 44859184-44859184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760687087 Chr.17: 44859035-44859036 on Build GRCh38
rs756297416 Chr.17: 44859043-44859043 on Build GRCh38
rs558069278 Chr.17: 44859076-44859076 on Build GRCh38
rs749556594 Chr.17: 44859063-44859063 on Build GRCh38
rs573434205 Chr.17: 44859203-44859203 on Build GRCh38
rs755245049 Chr.17: 44859054-44859054 on Build GRCh38
rs764767656 Chr.17: 44859166-44859166 on Build GRCh38
rs779023254 Chr.17: 44859230-44859230 on Build GRCh38
rs371715842 Chr.17: 44859193-44859193 on Build GRCh38
rs767886485 Chr.17: 44859179-44859179 on Build GRCh38
rs903725517 Chr.17: 44859181-44859181 on Build GRCh38
rs762110282 Chr.17: 44859170-44859170 on Build GRCh38
rs375672224 Chr.17: 44859047-44859047 on Build GRCh38
rs368015393 Chr.17: 44859053-44859053 on Build GRCh38
rs771850477 Chr.17: 44859067-44859067 on Build GRCh38
rs746880938 Chr.17: 44859110-44859110 on Build GRCh38
rs759253573 Chr.17: 44859164-44859164 on Build GRCh38
rs753935067 Chr.17: 44859204-44859204 on Build GRCh38
rs551580178 Chr.17: 44859069-44859069 on Build GRCh38
rs770443208 Chr.17: 44859122-44859122 on Build GRCh38
rs748361920 Chr.17: 44859231-44859231 on Build GRCh38
rs387906879 Chr.17: 44859132-44859132 on Build GRCh38
rs374713895 Chr.17: 44859195-44859195 on Build GRCh38
rs917916890 Chr.17: 44859163-44859163 on Build GRCh38
rs755298497 Chr.17: 44859222-44859222 on Build GRCh38
rs945261338 Chr.17: 44859135-44859135 on Build GRCh38
rs774829944 Chr.17: 44859167-44859167 on Build GRCh38
rs776105564 Chr.17: 44859148-44859148 on Build GRCh38
rs750897363 Chr.17: 44859038-44859038 on Build GRCh38
rs767891030 Chr.17: 44859032-44859032 on Build GRCh38
rs369215036 Chr.17: 44859062-44859062 on Build GRCh38

View Full Product Details