Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45830432-45830705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR1
Entrez Gene ID
1394
Gene Name
corticotropin releasing hormone receptor 1
Gene Aliases
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
UniGene
Hs.417628
Gene Chromosome Location
Chr.17: 45784280-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 45620329-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5095053 c.787A>T p.I263F Chr.17: 45830561-45830561 on Build GRCh38
4067113 c.652G>A p.V218M Chr.17: 45830513-45830513 on Build GRCh38
5985994 c.369C>G p.D123E Chr.17: 45830533-45830533 on Build GRCh38
4067112 c.644C>T p.T215I Chr.17: 45830505-45830505 on Build GRCh38
5055531 c.766C>T p.R256C Chr.17: 45830540-45830540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535553518 Chr.17: 45830507-45830507 on Build GRCh38
rs753364632 Chr.17: 45830615-45830615 on Build GRCh38
rs201094472 Chr.17: 45830580-45830580 on Build GRCh38
rs1048241138 Chr.17: 45830684-45830684 on Build GRCh38
rs746103309 Chr.17: 45830591-45830591 on Build GRCh38
rs943680561 Chr.17: 45830640-45830640 on Build GRCh38
rs369145044 Chr.17: 45830515-45830515 on Build GRCh38
rs531782343 Chr.17: 45830674-45830674 on Build GRCh38
rs16940668 Chr.17: 45830600-45830600 on Build GRCh38
rs918480673 Chr.17: 45830649-45830649 on Build GRCh38
rs760429758 Chr.17: 45830612-45830612 on Build GRCh38
rs868162436 Chr.17: 45830486-45830486 on Build GRCh38
rs765769414 Chr.17: 45830613-45830613 on Build GRCh38
rs377721543 Chr.17: 45830555-45830555 on Build GRCh38
rs757559668 Chr.17: 45830622-45830622 on Build GRCh38
rs767429408 Chr.17: 45830608-45830608 on Build GRCh38
rs776892398 Chr.17: 45830505-45830505 on Build GRCh38
rs562668116 Chr.17: 45830647-45830647 on Build GRCh38
rs779287406 Chr.17: 45830458-45830458 on Build GRCh38
rs769712683 Chr.17: 45830592-45830592 on Build GRCh38
rs564362344 Chr.17: 45830597-45830597 on Build GRCh38
rs543089732 Chr.17: 45830624-45830624 on Build GRCh38
rs779632730 Chr.17: 45830551-45830551 on Build GRCh38
rs778224446 Chr.17: 45830500-45830500 on Build GRCh38
rs895009803 Chr.17: 45830685-45830685 on Build GRCh38
rs755743910 Chr.17: 45830452-45830452 on Build GRCh38
rs1000182987 Chr.17: 45830566-45830566 on Build GRCh38
rs529332664 Chr.17: 45830623-45830623 on Build GRCh38
rs575488369 Chr.17: 45830534-45830534 on Build GRCh38
rs774745972 Chr.17: 45830467-45830469 on Build GRCh38
rs769867467 Chr.17: 45830512-45830512 on Build GRCh38
rs370336999 Chr.17: 45830540-45830540 on Build GRCh38
rs931238151 Chr.17: 45830653-45830653 on Build GRCh38
rs755621588 Chr.17: 45830541-45830541 on Build GRCh38
rs200096809 Chr.17: 45830618-45830618 on Build GRCh38
rs540215107 Chr.17: 45830616-45830616 on Build GRCh38
rs781324699 Chr.17: 45830585-45830585 on Build GRCh38
rs772269435 Chr.17: 45830482-45830482 on Build GRCh38
rs775232888 Chr.17: 45830513-45830513 on Build GRCh38
rs16940665 Chr.17: 45830530-45830530 on Build GRCh38
rs992144291 Chr.17: 45830598-45830598 on Build GRCh38
rs369593816 Chr.17: 45830675-45830675 on Build GRCh38
rs762985262 Chr.17: 45830593-45830593 on Build GRCh38
rs771347065 Chr.17: 45830584-45830584 on Build GRCh38
rs376604761 Chr.17: 45830535-45830535 on Build GRCh38
rs748697265 Chr.17: 45830474-45830474 on Build GRCh38
rs544464954 Chr.17: 45830552-45830552 on Build GRCh38
rs150922527 Chr.17: 45830659-45830659 on Build GRCh38
rs768618052 Chr.17: 45830594-45830594 on Build GRCh38
rs935832459 Chr.17: 45830627-45830627 on Build GRCh38
rs771116141 Chr.17: 45830503-45830503 on Build GRCh38
rs750041935 Chr.17: 45830609-45830609 on Build GRCh38
rs368177064 Chr.17: 45830485-45830485 on Build GRCh38
rs778189231 Chr.17: 45830556-45830556 on Build GRCh38
rs767265946 Chr.17: 45830539-45830539 on Build GRCh38
rs895209460 Chr.17: 45830470-45830470 on Build GRCh38

View Full Product Details