Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85110247-85110502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92B
Entrez Gene ID
339145
Gene Name
family with sequence similarity 92 member B
Gene Aliases
-
UniGene
Hs.125713
Gene Chromosome Location
Chr.16: 85098352-85112508 on Build GRCh38
Transcripts
NM_198491.2 XM_005255931.2 XM_011523063.1 XM_017023198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830841 c.127C>T p.R43W Chr.16: 85110354-85110354 on Build GRCh38
6555960 c.193C>A p.R65R Chr.16: 85110288-85110288 on Build GRCh38
4808873 c.46A>G p.N16D Chr.16: 85110435-85110435 on Build GRCh38
6523073 c.63C>T p.T21T Chr.16: 85110418-85110418 on Build GRCh38
197317 c.116C>T p.T39M Chr.16: 85110365-85110365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773751395 Chr.16: 85110386-85110386 on Build GRCh38
rs771669601 Chr.16: 85110304-85110304 on Build GRCh38
rs753996502 Chr.16: 85110360-85110360 on Build GRCh38
rs199531863 Chr.16: 85110334-85110334 on Build GRCh38
rs374566330 Chr.16: 85110295-85110295 on Build GRCh38
rs375570292 Chr.16: 85110275-85110275 on Build GRCh38
rs759100677 Chr.16: 85110405-85110405 on Build GRCh38
rs546799870 Chr.16: 85110278-85110278 on Build GRCh38
rs772753183 Chr.16: 85110439-85110439 on Build GRCh38
rs1041867817 Chr.16: 85110296-85110296 on Build GRCh38
rs377518196 Chr.16: 85110422-85110422 on Build GRCh38
rs188484129 Chr.16: 85110285-85110285 on Build GRCh38
rs768163657 Chr.16: 85110380-85110380 on Build GRCh38
rs750085302 Chr.16: 85110481-85110481 on Build GRCh38
rs562127957 Chr.16: 85110340-85110340 on Build GRCh38
rs1004722397 Chr.16: 85110346-85110346 on Build GRCh38
rs772868767 Chr.16: 85110307-85110307 on Build GRCh38
rs372625485 Chr.16: 85110270-85110270 on Build GRCh38
rs763349577 Chr.16: 85110319-85110319 on Build GRCh38
rs35834733 Chr.16: 85110282-85110282 on Build GRCh38
rs9934891 Chr.16: 85110417-85110417 on Build GRCh38
rs1050988910 Chr.16: 85110479-85110479 on Build GRCh38
rs760784798 Chr.16: 85110388-85110388 on Build GRCh38
rs201562054 Chr.16: 85110301-85110301 on Build GRCh38
rs887852586 Chr.16: 85110289-85110289 on Build GRCh38
rs1017051416 Chr.16: 85110349-85110349 on Build GRCh38
rs775012665 Chr.16: 85110381-85110381 on Build GRCh38
rs763570040 Chr.16: 85110268-85110268 on Build GRCh38
rs182096113 Chr.16: 85110303-85110303 on Build GRCh38
rs149617066 Chr.16: 85110297-85110297 on Build GRCh38
rs140432540 Chr.16: 85110272-85110272 on Build GRCh38
rs138142092 Chr.16: 85110364-85110364 on Build GRCh38
rs766128769 Chr.16: 85110368-85110368 on Build GRCh38
rs374760302 Chr.16: 85110442-85110442 on Build GRCh38
rs751394451 Chr.16: 85110411-85110411 on Build GRCh38
rs767273445 Chr.16: 85110477-85110477 on Build GRCh38
rs761128607 Chr.16: 85110450-85110450 on Build GRCh38
rs370185701 Chr.16: 85110363-85110363 on Build GRCh38
rs750472893 Chr.16: 85110294-85110294 on Build GRCh38
rs780903491 Chr.16: 85110418-85110418 on Build GRCh38
rs199902827 Chr.16: 85110407-85110407 on Build GRCh38
rs758180795 Chr.16: 85110371-85110371 on Build GRCh38
rs372890171 Chr.16: 85110470-85110470 on Build GRCh38
rs776922191 Chr.16: 85110454-85110454 on Build GRCh38
rs757090252 Chr.16: 85110474-85110474 on Build GRCh38
rs143063631 Chr.16: 85110404-85110404 on Build GRCh38
rs900059030 Chr.16: 85110413-85110413 on Build GRCh38
rs773398398 Chr.16: 85110448-85110448 on Build GRCh38
rs769416097 Chr.16: 85110423-85110423 on Build GRCh38
rs1004244141 Chr.16: 85110455-85110455 on Build GRCh38
rs752629671 Chr.16: 85110401-85110401 on Build GRCh38
rs748006465 Chr.16: 85110359-85110359 on Build GRCh38
rs765272850 Chr.16: 85110461-85110461 on Build GRCh38
rs766643279 Chr.16: 85110391-85110391 on Build GRCh38
rs759578773 Chr.16: 85110344-85110345 on Build GRCh38
rs764052710 Chr.16: 85110471-85110471 on Build GRCh38
rs780520944 Chr.16: 85110374-85110374 on Build GRCh38
rs767873697 Chr.16: 85110287-85110287 on Build GRCh38
rs146980409 Chr.16: 85110343-85110343 on Build GRCh38
rs948075322 Chr.16: 85110387-85110387 on Build GRCh38
rs764341130 Chr.16: 85110273-85110273 on Build GRCh38
rs141444139 Chr.16: 85110318-85110318 on Build GRCh38
rs765362750 Chr.16: 85110397-85110397 on Build GRCh38
rs552316888 Chr.16: 85110457-85110457 on Build GRCh38
rs762183805 Chr.16: 85110291-85110291 on Build GRCh38
rs561364904 Chr.16: 85110375-85110375 on Build GRCh38
rs146138709 Chr.16: 85110472-85110472 on Build GRCh38
rs750660848 Chr.16: 85110344-85110344 on Build GRCh38
rs201987292 Chr.16: 85110365-85110365 on Build GRCh38
rs746725470 Chr.16: 85110308-85110308 on Build GRCh38
rs770989501 Chr.16: 85110451-85110451 on Build GRCh38
rs777615366 Chr.16: 85110372-85110372 on Build GRCh38
rs530041314 Chr.16: 85110384-85110384 on Build GRCh38
rs202030185 Chr.16: 85110354-85110354 on Build GRCh38
rs764685135 Chr.16: 85110322-85110322 on Build GRCh38
rs774816773 Chr.16: 85110333-85110333 on Build GRCh38
rs772559148 Chr.16: 85110385-85110385 on Build GRCh38
rs761023982 Chr.16: 85110353-85110353 on Build GRCh38
rs145597678 Chr.16: 85110298-85110298 on Build GRCh38
rs143497220 Chr.16: 85110373-85110373 on Build GRCh38
rs775852923 Chr.16: 85110267-85110267 on Build GRCh38
rs755871002 Chr.16: 85110482-85110482 on Build GRCh38
rs779776033 Chr.16: 85110433-85110433 on Build GRCh38
rs748633466 Chr.16: 85110435-85110435 on Build GRCh38
rs186747840 Chr.16: 85110309-85110309 on Build GRCh38
rs753091695 Chr.16: 85110392-85110392 on Build GRCh38

View Full Product Details