Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39840275-39840548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBL
Entrez Gene ID
2091
Gene Name
fibrillarin
Gene Aliases
FIB, FLRN, RNU3IP1
UniGene
Hs.299002
Gene Chromosome Location
Chr.19: 39834453-39846414 on Build GRCh38
Transcripts
NM_001436.3 XM_005258651.2 XM_011526623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266523 c.206A>T p.N69I Chr.19: 39840491-39840491 on Build GRCh38
3990016 c.336C>T p.V112V Chr.19: 39840275-39840275 on Build GRCh38
6571729 c.245_246insTC p.G83fs*5 Chr.19: 39840451-39840452 on Build GRCh38
6456363 c.197C>T p.S66F Chr.19: 39840500-39840500 on Build GRCh38
1750890 c.281A>G p.E94G Chr.19: 39840416-39840416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146399599 Chr.19: 39840452-39840452 on Build GRCh38
rs369326013 Chr.19: 39840427-39840427 on Build GRCh38
rs142991383 Chr.19: 39840482-39840482 on Build GRCh38
rs759364850 Chr.19: 39840352-39840352 on Build GRCh38
rs141906890 Chr.19: 39840451-39840451 on Build GRCh38
rs529542338 Chr.19: 39840338-39840338 on Build GRCh38
rs139739777 Chr.19: 39840477-39840477 on Build GRCh38
rs150661315 Chr.19: 39840488-39840488 on Build GRCh38
rs756729596 Chr.19: 39840489-39840489 on Build GRCh38
rs767999959 Chr.19: 39840485-39840485 on Build GRCh38
rs377289250 Chr.19: 39840379-39840379 on Build GRCh38
rs1029303047 Chr.19: 39840457-39840457 on Build GRCh38
rs776790722 Chr.19: 39840472-39840472 on Build GRCh38
rs770600761 Chr.19: 39840343-39840343 on Build GRCh38
rs745547248 Chr.19: 39840440-39840440 on Build GRCh38
rs370007580 Chr.19: 39840350-39840350 on Build GRCh38
rs201702222 Chr.19: 39840390-39840390 on Build GRCh38
rs377102927 Chr.19: 39840393-39840393 on Build GRCh38
rs534041209 Chr.19: 39840493-39840493 on Build GRCh38
rs778623138 Chr.19: 39840437-39840437 on Build GRCh38
rs201617067 Chr.19: 39840428-39840428 on Build GRCh38
rs111746297 Chr.19: 39840520-39840520 on Build GRCh38
rs761969290 Chr.19: 39840475-39840475 on Build GRCh38
rs200224908 Chr.19: 39840433-39840433 on Build GRCh38
rs755409839 Chr.19: 39840483-39840483 on Build GRCh38
rs747533141 Chr.19: 39840295-39840295 on Build GRCh38
rs878909159 Chr.19: 39840409-39840409 on Build GRCh38
rs771829791 Chr.19: 39840358-39840358 on Build GRCh38
rs775468086 Chr.19: 39840372-39840372 on Build GRCh38
rs770488310 Chr.19: 39840435-39840435 on Build GRCh38
rs771881501 Chr.19: 39840442-39840442 on Build GRCh38
rs765475178 Chr.19: 39840476-39840476 on Build GRCh38
rs113832786 Chr.19: 39840518-39840518 on Build GRCh38
rs779833804 Chr.19: 39840510-39840510 on Build GRCh38
rs769261583 Chr.19: 39840298-39840298 on Build GRCh38
rs758022467 Chr.19: 39840503-39840503 on Build GRCh38
rs745479581 Chr.19: 39840499-39840499 on Build GRCh38
rs757852054 Chr.19: 39840445-39840453 on Build GRCh38
rs200890919 Chr.19: 39840391-39840391 on Build GRCh38
rs777313281 Chr.19: 39840320-39840320 on Build GRCh38
rs879085400 Chr.19: 39840410-39840410 on Build GRCh38
rs550186747 Chr.19: 39840380-39840380 on Build GRCh38
rs752353507 Chr.19: 39840378-39840379 on Build GRCh38
rs760581781 Chr.19: 39840376-39840376 on Build GRCh38
rs2302113 Chr.19: 39840478-39840478 on Build GRCh38
rs768721519 Chr.19: 39840460-39840460 on Build GRCh38

View Full Product Details