Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43040387-43040634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
UniGene
Hs.520136
Gene Chromosome Location
Chr.6: 43037617-43053950 on Build GRCh38
Transcripts
NM_001168370.1 NM_014780.4 XM_005249503.2 XM_006715285.1 XM_011515019.2 XM_011515020.2 XM_011515021.1 XM_017011533.1 XM_017011534.1 XM_017011535.1 XM_017011536.1 XM_017011537.1 XM_017011538.1 XM_017011539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278695 c.4198C>T p.R1400C Chr.6: 43040607-43040607 on Build GRCh38
5425417 c.3993G>A p.L1331L Chr.6: 43040560-43040560 on Build GRCh38
4906343 c.4225C>T p.Q1409* Chr.6: 43040580-43040580 on Build GRCh38
4651582 c.4195C>T p.Q1399* Chr.6: 43040610-43040610 on Build GRCh38
5935104 c.4030C>T p.L1344F Chr.6: 43040420-43040420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770815047 Chr.6: 43040595-43040595 on Build GRCh38
rs777870519 Chr.6: 43040575-43040575 on Build GRCh38
rs762448194 Chr.6: 43040435-43040435 on Build GRCh38
rs750745627 Chr.6: 43040534-43040534 on Build GRCh38
rs376360115 Chr.6: 43040460-43040460 on Build GRCh38
rs762248583 Chr.6: 43040532-43040532 on Build GRCh38
rs151030955 Chr.6: 43040558-43040558 on Build GRCh38
rs750439460 Chr.6: 43040495-43040495 on Build GRCh38
rs759270786 Chr.6: 43040521-43040521 on Build GRCh38
rs141784862 Chr.6: 43040479-43040479 on Build GRCh38
rs767854529 Chr.6: 43040436-43040436 on Build GRCh38
rs548538046 Chr.6: 43040420-43040420 on Build GRCh38
rs142830307 Chr.6: 43040550-43040550 on Build GRCh38
rs1004584584 Chr.6: 43040587-43040587 on Build GRCh38
rs1041384876 Chr.6: 43040424-43040424 on Build GRCh38
rs752831085 Chr.6: 43040566-43040566 on Build GRCh38
rs1048929663 Chr.6: 43040559-43040559 on Build GRCh38
rs976657977 Chr.6: 43040450-43040450 on Build GRCh38
rs1012731965 Chr.6: 43040545-43040545 on Build GRCh38
rs776120876 Chr.6: 43040503-43040503 on Build GRCh38
rs746836526 Chr.6: 43040496-43040496 on Build GRCh38
rs963922002 Chr.6: 43040429-43040429 on Build GRCh38
rs751976526 Chr.6: 43040434-43040434 on Build GRCh38
rs139611086 Chr.6: 43040544-43040544 on Build GRCh38
rs750798764 Chr.6: 43040440-43040440 on Build GRCh38
rs758375607 Chr.6: 43040574-43040574 on Build GRCh38
rs776163035 Chr.6: 43040407-43040407 on Build GRCh38
rs769180724 Chr.6: 43040523-43040523 on Build GRCh38
rs370901005 Chr.6: 43040478-43040478 on Build GRCh38
rs775122122 Chr.6: 43040527-43040527 on Build GRCh38
rs769499040 Chr.6: 43040607-43040607 on Build GRCh38
rs922465157 Chr.6: 43040465-43040465 on Build GRCh38
rs768016103 Chr.6: 43040533-43040533 on Build GRCh38
rs781105907 Chr.6: 43040596-43040596 on Build GRCh38
rs777647658 Chr.6: 43040486-43040486 on Build GRCh38
rs745491935 Chr.6: 43040601-43040601 on Build GRCh38
rs746988149 Chr.6: 43040584-43040584 on Build GRCh38
rs2273917 Chr.6: 43040560-43040560 on Build GRCh38
rs758986941 Chr.6: 43040415-43040415 on Build GRCh38
rs780278402 Chr.6: 43040466-43040466 on Build GRCh38
rs748025697 Chr.6: 43040480-43040480 on Build GRCh38
rs571067327 Chr.6: 43040481-43040481 on Build GRCh38
rs375477525 Chr.6: 43040488-43040488 on Build GRCh38

View Full Product Details