Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34761993-34762224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB4
Entrez Gene ID
127534
Gene Name
gap junction protein beta 4
Gene Aliases
CX30.3, EKV
UniGene
Hs.351203
Gene Chromosome Location
Chr.1: 34759741-34763724 on Build GRCh38
Transcripts
NM_153212.2 XM_011540679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488824 c.739G>A p.D247N Chr.1: 34761993-34761993 on Build GRCh38
4402341 c.770C>T p.S257L Chr.1: 34762024-34762024 on Build GRCh38
908575 c.746A>C p.N249T Chr.1: 34762000-34762000 on Build GRCh38
5420351 c.792G>T p.G264G Chr.1: 34762046-34762046 on Build GRCh38
6671228 c.764C>T p.A255V Chr.1: 34762018-34762018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182726132 Chr.1: 34762098-34762098 on Build GRCh38
rs144672584 Chr.1: 34762185-34762185 on Build GRCh38
rs980510112 Chr.1: 34762171-34762171 on Build GRCh38
rs778589244 Chr.1: 34762118-34762118 on Build GRCh38
rs1015492770 Chr.1: 34762110-34762110 on Build GRCh38
rs745891365 Chr.1: 34762093-34762094 on Build GRCh38
rs1055922124 Chr.1: 34762087-34762087 on Build GRCh38
rs532786755 Chr.1: 34762198-34762198 on Build GRCh38
rs763471752 Chr.1: 34762077-34762077 on Build GRCh38
rs777035236 Chr.1: 34762063-34762063 on Build GRCh38
rs913723524 Chr.1: 34762203-34762203 on Build GRCh38
rs968999679 Chr.1: 34762167-34762167 on Build GRCh38
rs765840735 Chr.1: 34762082-34762082 on Build GRCh38
rs538347700 Chr.1: 34762039-34762039 on Build GRCh38
rs757149751 Chr.1: 34762105-34762105 on Build GRCh38
rs755931 Chr.1: 34762072-34762072 on Build GRCh38
rs775597117 Chr.1: 34762046-34762046 on Build GRCh38
rs772288539 Chr.1: 34762096-34762097 on Build GRCh38
rs749213359 Chr.1: 34762102-34762102 on Build GRCh38
rs199996022 Chr.1: 34762017-34762017 on Build GRCh38
rs754603988 Chr.1: 34762091-34762091 on Build GRCh38
rs901806045 Chr.1: 34762085-34762085 on Build GRCh38
rs376640390 Chr.1: 34762060-34762060 on Build GRCh38
rs780011671 Chr.1: 34762106-34762106 on Build GRCh38
rs1024643214 Chr.1: 34762161-34762161 on Build GRCh38
rs1006416307 Chr.1: 34762109-34762109 on Build GRCh38
rs879111187 Chr.1: 34762193-34762193 on Build GRCh38
rs186793796 Chr.1: 34762141-34762141 on Build GRCh38
rs771683313 Chr.1: 34762186-34762186 on Build GRCh38
rs148656277 Chr.1: 34762024-34762024 on Build GRCh38
rs932062714 Chr.1: 34762087-34762087 on Build GRCh38
rs751019283 Chr.1: 34762088-34762088 on Build GRCh38
rs556066067 Chr.1: 34762018-34762018 on Build GRCh38
rs762156674 Chr.1: 34762066-34762066 on Build GRCh38
rs777623272 Chr.1: 34762061-34762061 on Build GRCh38
rs1010697749 Chr.1: 34762022-34762022 on Build GRCh38
rs1017685855 Chr.1: 34762164-34762164 on Build GRCh38
rs765771714 Chr.1: 34762071-34762071 on Build GRCh38
rs756019816 Chr.1: 34762025-34762025 on Build GRCh38
rs951100124 Chr.1: 34762194-34762194 on Build GRCh38
rs999143340 Chr.1: 34762140-34762140 on Build GRCh38
rs1055116153 Chr.1: 34762153-34762153 on Build GRCh38
rs777849647 Chr.1: 34762027-34762027 on Build GRCh38
rs779022220 Chr.1: 34762086-34762087 on Build GRCh38
rs372744155 Chr.1: 34762093-34762093 on Build GRCh38
rs772066664 Chr.1: 34762045-34762045 on Build GRCh38
rs976388855 Chr.1: 34762034-34762034 on Build GRCh38
rs76306134 Chr.1: 34762115-34762115 on Build GRCh38

View Full Product Details