Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128694192-128694441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
UniGene
Hs.504281
Gene Chromosome Location
Chr.11: 128685263-128813267 on Build GRCh38
Transcripts
NM_001167681.2 NM_001271010.1 NM_001271012.1 NM_002017.4 XM_011542701.2 XM_011542702.1 XM_017017405.1 XM_017017406.1 NR_038908.1

Gene Symbol
SENCR
Entrez Gene ID
100507392
Gene Name
smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA
Gene Aliases
FLI1-AS1, lncRNA9
UniGene
Hs.657715
Gene Chromosome Location
Chr.11: 128691672-128696023 on Build GRCh38
Transcripts
NM_001167681.2 NM_001271010.1 NM_001271012.1 NM_002017.4 XM_011542701.2 XM_011542702.1 XM_017017405.1 XM_017017406.1 NR_038908.1

SNP Detail

SNP ID Chromosome Location
rs987033963 Chr.11: 128694403-128694403 on Build GRCh38
rs563839566 Chr.11: 128694346-128694346 on Build GRCh38
rs148819776 Chr.11: 128694362-128694362 on Build GRCh38
rs775206695 Chr.11: 128694314-128694314 on Build GRCh38
rs529350826 Chr.11: 128694349-128694349 on Build GRCh38
rs974527541 Chr.11: 128694341-128694341 on Build GRCh38
rs868682780 Chr.11: 128694361-128694361 on Build GRCh38
rs1033274744 Chr.11: 128694347-128694347 on Build GRCh38
rs766178380 Chr.11: 128694245-128694245 on Build GRCh38
rs746564927 Chr.11: 128694231-128694232 on Build GRCh38
rs760147027 Chr.11: 128694388-128694388 on Build GRCh38
rs765637863 Chr.11: 128694424-128694424 on Build GRCh38
rs112860445 Chr.11: 128694231-128694231 on Build GRCh38
rs369578409 Chr.11: 128694289-128694289 on Build GRCh38
rs773962272 Chr.11: 128694312-128694312 on Build GRCh38
rs752471336 Chr.11: 128694266-128694266 on Build GRCh38
rs984340834 Chr.11: 128694338-128694338 on Build GRCh38
rs748529525 Chr.11: 128694299-128694299 on Build GRCh38
rs773821278 Chr.11: 128694303-128694303 on Build GRCh38
rs770487665 Chr.11: 128694300-128694300 on Build GRCh38
rs966176013 Chr.11: 128694311-128694311 on Build GRCh38
rs780059235 Chr.11: 128694288-128694288 on Build GRCh38
rs1047214343 Chr.11: 128694236-128694236 on Build GRCh38
rs759119632 Chr.11: 128694313-128694313 on Build GRCh38
rs377696295 Chr.11: 128694309-128694309 on Build GRCh38
rs767250889 Chr.11: 128694265-128694265 on Build GRCh38
rs768231266 Chr.11: 128694302-128694328 on Build GRCh38
rs550022199 Chr.11: 128694326-128694326 on Build GRCh38
rs750570259 Chr.11: 128694286-128694286 on Build GRCh38
rs769220392 Chr.11: 128694399-128694399 on Build GRCh38
rs776178042 Chr.11: 128694310-128694311 on Build GRCh38
rs1031551701 Chr.11: 128694242-128694242 on Build GRCh38
rs774127474 Chr.11: 128694247-128694247 on Build GRCh38
rs1037979628 Chr.11: 128694210-128694210 on Build GRCh38
rs940050834 Chr.11: 128694372-128694372 on Build GRCh38
rs573220409 Chr.11: 128694284-128694284 on Build GRCh38
rs756020021 Chr.11: 128694272-128694272 on Build GRCh38
rs762545912 Chr.11: 128694235-128694235 on Build GRCh38
rs748604139 Chr.11: 128694227-128694227 on Build GRCh38
rs759182658 Chr.11: 128694255-128694255 on Build GRCh38
rs955904174 Chr.11: 128694267-128694267 on Build GRCh38
rs564476686 Chr.11: 128694307-128694307 on Build GRCh38
rs919567291 Chr.11: 128694211-128694211 on Build GRCh38
rs758449121 Chr.11: 128694287-128694287 on Build GRCh38
rs777111125 Chr.11: 128694411-128694411 on Build GRCh38
rs376557878 Chr.11: 128694230-128694230 on Build GRCh38
rs921587437 Chr.11: 128694345-128694345 on Build GRCh38
rs565927768 Chr.11: 128694396-128694396 on Build GRCh38
rs771580215 Chr.11: 128694385-128694385 on Build GRCh38
rs1020752025 Chr.11: 128694394-128694394 on Build GRCh38
rs1019149036 Chr.11: 128694293-128694293 on Build GRCh38
rs1057132459 Chr.11: 128694377-128694377 on Build GRCh38
rs762248398 Chr.11: 128694416-128694416 on Build GRCh38
rs895914871 Chr.11: 128694380-128694380 on Build GRCh38
rs201691053 Chr.11: 128694294-128694294 on Build GRCh38
rs948820450 Chr.11: 128694390-128694390 on Build GRCh38
rs775115826 Chr.11: 128694386-128694386 on Build GRCh38
rs772912959 Chr.11: 128694233-128694233 on Build GRCh38

View Full Product Details