Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66000154-66000415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AD
Entrez Gene ID
84285
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
haponin
UniGene
Hs.425178
Gene Chromosome Location
Chr.11: 65996545-66002217 on Build GRCh38
Transcripts
NM_001242481.1 NM_001242482.1 NM_001242483.1 NM_001242484.1 NM_001242485.1 NM_001242486.1 NM_032325.3 XM_017018412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035605 c.85A>G p.R29G Chr.11: 66000305-66000305 on Build GRCh38
429625 c.13A>G p.T5A Chr.11: 66000377-66000377 on Build GRCh38
5444833 c.18G>C p.K6N Chr.11: 66000372-66000372 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373542164 Chr.11: 66000293-66000293 on Build GRCh38
rs773701677 Chr.11: 66000363-66000363 on Build GRCh38
rs780399408 Chr.11: 66000285-66000285 on Build GRCh38
rs1051037601 Chr.11: 66000240-66000240 on Build GRCh38
rs377089346 Chr.11: 66000213-66000213 on Build GRCh38
rs752888050 Chr.11: 66000389-66000389 on Build GRCh38
rs149773778 Chr.11: 66000381-66000381 on Build GRCh38
rs750915943 Chr.11: 66000270-66000270 on Build GRCh38
rs761183142 Chr.11: 66000366-66000366 on Build GRCh38
rs749157885 Chr.11: 66000263-66000263 on Build GRCh38
rs762702705 Chr.11: 66000207-66000207 on Build GRCh38
rs574841858 Chr.11: 66000221-66000221 on Build GRCh38
rs776473849 Chr.11: 66000181-66000181 on Build GRCh38
rs978412339 Chr.11: 66000359-66000359 on Build GRCh38
rs758458281 Chr.11: 66000317-66000317 on Build GRCh38
rs375615308 Chr.11: 66000204-66000204 on Build GRCh38
rs905224640 Chr.11: 66000223-66000223 on Build GRCh38
rs929777409 Chr.11: 66000236-66000236 on Build GRCh38
rs369633698 Chr.11: 66000294-66000294 on Build GRCh38
rs769649602 Chr.11: 66000327-66000327 on Build GRCh38
rs376749430 Chr.11: 66000180-66000180 on Build GRCh38
rs749157947 Chr.11: 66000332-66000332 on Build GRCh38
rs754198174 Chr.11: 66000374-66000374 on Build GRCh38
rs754000096 Chr.11: 66000289-66000289 on Build GRCh38
rs372674906 Chr.11: 66000192-66000192 on Build GRCh38
rs773836534 Chr.11: 66000193-66000193 on Build GRCh38
rs372901719 Chr.11: 66000338-66000338 on Build GRCh38
rs541380914 Chr.11: 66000217-66000217 on Build GRCh38
rs376933677 Chr.11: 66000371-66000371 on Build GRCh38
rs745828894 Chr.11: 66000324-66000324 on Build GRCh38
rs763894420 Chr.11: 66000210-66000210 on Build GRCh38
rs201150583 Chr.11: 66000382-66000382 on Build GRCh38
rs140918998 Chr.11: 66000246-66000246 on Build GRCh38
rs1006784913 Chr.11: 66000255-66000255 on Build GRCh38
rs747445566 Chr.11: 66000256-66000256 on Build GRCh38
rs756429909 Chr.11: 66000275-66000275 on Build GRCh38
rs751073532 Chr.11: 66000211-66000211 on Build GRCh38
rs1022879038 Chr.11: 66000222-66000222 on Build GRCh38
rs368761728 Chr.11: 66000186-66000186 on Build GRCh38
rs17849919 Chr.11: 66000323-66000323 on Build GRCh38
rs761336884 Chr.11: 66000351-66000351 on Build GRCh38
rs775423191 Chr.11: 66000260-66000260 on Build GRCh38
rs775442481 Chr.11: 66000328-66000328 on Build GRCh38
rs773921780 Chr.11: 66000343-66000343 on Build GRCh38
rs148239309 Chr.11: 66000361-66000361 on Build GRCh38
rs1016303491 Chr.11: 66000250-66000250 on Build GRCh38

View Full Product Details