Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 844877-845130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110A
Entrez Gene ID
83541
Gene Name
family with sequence similarity 110 member A
Gene Aliases
C20orf55, F10, bA371L19.3
UniGene
Hs.574822
Gene Chromosome Location
Chr.20: 833697-846279 on Build GRCh38
Transcripts
NM_001042353.2 NM_001289145.1 NM_001289146.1 NM_001289147.1 NM_031424.5 NM_207121.4 XM_011529381.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226762 c.185C>T p.T62I Chr.20: 844989-844989 on Build GRCh38
3693680 c.260C>T p.T87M Chr.20: 845064-845064 on Build GRCh38
5676176 c.120G>T p.P40P Chr.20: 844924-844924 on Build GRCh38
1029304 c.211C>T p.L71L Chr.20: 845015-845015 on Build GRCh38
1743293 c.276C>T p.A92A Chr.20: 845080-845080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567300552 Chr.20: 845070-845070 on Build GRCh38
rs764162128 Chr.20: 844957-844957 on Build GRCh38
rs139752871 Chr.20: 844964-844964 on Build GRCh38
rs8124809 Chr.20: 844936-844936 on Build GRCh38
rs537185607 Chr.20: 845029-845029 on Build GRCh38
rs980222803 Chr.20: 845019-845019 on Build GRCh38
rs755770947 Chr.20: 844999-844999 on Build GRCh38
rs752710189 Chr.20: 845091-845091 on Build GRCh38
rs150649866 Chr.20: 844940-844940 on Build GRCh38
rs867123388 Chr.20: 844991-844991 on Build GRCh38
rs914093059 Chr.20: 845089-845089 on Build GRCh38
rs779625752 Chr.20: 845002-845002 on Build GRCh38
rs771289212 Chr.20: 845052-845052 on Build GRCh38
rs762106069 Chr.20: 844914-844914 on Build GRCh38
rs868087764 Chr.20: 844982-844982 on Build GRCh38
rs973229969 Chr.20: 844930-844930 on Build GRCh38
rs571318815 Chr.20: 844962-844962 on Build GRCh38
rs769153392 Chr.20: 844910-844910 on Build GRCh38
rs374393104 Chr.20: 845100-845100 on Build GRCh38
rs374261858 Chr.20: 845033-845033 on Build GRCh38
rs6140402 Chr.20: 844913-844913 on Build GRCh38
rs144349814 Chr.20: 844915-844915 on Build GRCh38
rs773418881 Chr.20: 844924-844924 on Build GRCh38
rs765006821 Chr.20: 845081-845081 on Build GRCh38
rs748820460 Chr.20: 845023-845023 on Build GRCh38
rs371582845 Chr.20: 845051-845051 on Build GRCh38
rs765443318 Chr.20: 845076-845076 on Build GRCh38
rs750248681 Chr.20: 845110-845110 on Build GRCh38
rs866342248 Chr.20: 845059-845059 on Build GRCh38
rs757123863 Chr.20: 844985-844985 on Build GRCh38
rs762707860 Chr.20: 845060-845061 on Build GRCh38
rs750183788 Chr.20: 844989-844989 on Build GRCh38
rs772444407 Chr.20: 845048-845048 on Build GRCh38
rs781102126 Chr.20: 844987-844987 on Build GRCh38
rs761026151 Chr.20: 844934-844934 on Build GRCh38
rs368013856 Chr.20: 845049-845049 on Build GRCh38
rs759716075 Chr.20: 845075-845075 on Build GRCh38
rs966918025 Chr.20: 845008-845008 on Build GRCh38
rs538094 Chr.20: 845035-845035 on Build GRCh38
rs753982592 Chr.20: 844937-844937 on Build GRCh38
rs776940535 Chr.20: 845072-845072 on Build GRCh38

View Full Product Details