Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32228986-32229257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3I
Entrez Gene ID
8668
Gene Name
eukaryotic translation initiation factor 3 subunit I
Gene Aliases
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36
UniGene
Hs.530096
Gene Chromosome Location
Chr.1: 32221584-32231604 on Build GRCh38
Transcripts
NM_003757.3 XM_017002671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991653 c.803+4_803+6delAAG p.? Chr.1: 32229212-32229214 on Build GRCh38
5425319 c.803+10A>G p.? Chr.1: 32229218-32229218 on Build GRCh38
5416939 c.776C>T p.S259F Chr.1: 32229181-32229181 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934191548 Chr.1: 32229013-32229013 on Build GRCh38
rs182451914 Chr.1: 32229072-32229072 on Build GRCh38
rs571123805 Chr.1: 32229011-32229011 on Build GRCh38
rs753926762 Chr.1: 32229052-32229052 on Build GRCh38
rs931825624 Chr.1: 32229211-32229211 on Build GRCh38
rs761131050 Chr.1: 32229036-32229036 on Build GRCh38
rs781036679 Chr.1: 32229124-32229124 on Build GRCh38
rs1033248685 Chr.1: 32229035-32229035 on Build GRCh38
rs140552630 Chr.1: 32229043-32229043 on Build GRCh38
rs762515370 Chr.1: 32229102-32229102 on Build GRCh38
rs574786196 Chr.1: 32229048-32229048 on Build GRCh38
rs765751405 Chr.1: 32229145-32229146 on Build GRCh38
rs765892244 Chr.1: 32229099-32229099 on Build GRCh38
rs756058298 Chr.1: 32229159-32229159 on Build GRCh38
rs758414558 Chr.1: 32229203-32229203 on Build GRCh38
rs777645990 Chr.1: 32229161-32229161 on Build GRCh38
rs751025103 Chr.1: 32229211-32229213 on Build GRCh38
rs370375265 Chr.1: 32229121-32229121 on Build GRCh38
rs766012759 Chr.1: 32229107-32229107 on Build GRCh38
rs545905686 Chr.1: 32229188-32229188 on Build GRCh38
rs764680777 Chr.1: 32229042-32229042 on Build GRCh38
rs762364711 Chr.1: 32229096-32229096 on Build GRCh38
rs754476616 Chr.1: 32229212-32229214 on Build GRCh38
rs762244261 Chr.1: 32229119-32229121 on Build GRCh38
rs752493728 Chr.1: 32229143-32229143 on Build GRCh38
rs751203354 Chr.1: 32229120-32229120 on Build GRCh38
rs72666792 Chr.1: 32229218-32229218 on Build GRCh38
rs373646065 Chr.1: 32229093-32229093 on Build GRCh38
rs749244704 Chr.1: 32229197-32229197 on Build GRCh38
rs1033502006 Chr.1: 32229138-32229138 on Build GRCh38
rs200360014 Chr.1: 32229101-32229101 on Build GRCh38
rs970646499 Chr.1: 32229007-32229007 on Build GRCh38

View Full Product Details