Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19504706-19504979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDE1
Entrez Gene ID
51573
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
363E6.2, MIR16
UniGene
Hs.512607
Gene Chromosome Location
Chr.16: 19501693-19522145 on Build GRCh38
Transcripts
NM_001324066.1 NM_001324067.1 NM_016641.3 NR_136689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3506740 c.848C>T p.P283L Chr.16: 19504881-19504881 on Build GRCh38
5631794 c.754A>G p.M252V Chr.16: 19504975-19504975 on Build GRCh38
1742119 c.777G>C p.W259C Chr.16: 19504952-19504952 on Build GRCh38
6484247 c.759G>A p.M253I Chr.16: 19504970-19504970 on Build GRCh38
40884 c.769C>T p.L257F Chr.16: 19504960-19504960 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780579441 Chr.16: 19504876-19504877 on Build GRCh38
rs766052454 Chr.16: 19504847-19504847 on Build GRCh38
rs554574797 Chr.16: 19504816-19504816 on Build GRCh38
rs953827172 Chr.16: 19504836-19504836 on Build GRCh38
rs140559686 Chr.16: 19504799-19504799 on Build GRCh38
rs751165180 Chr.16: 19504850-19504850 on Build GRCh38
rs762672600 Chr.16: 19504845-19504845 on Build GRCh38
rs901181962 Chr.16: 19504812-19504812 on Build GRCh38
rs757643915 Chr.16: 19504929-19504929 on Build GRCh38
rs998334154 Chr.16: 19504806-19504806 on Build GRCh38
rs200268131 Chr.16: 19504899-19504899 on Build GRCh38
rs765003403 Chr.16: 19504839-19504839 on Build GRCh38
rs750924151 Chr.16: 19504861-19504862 on Build GRCh38
rs1047424277 Chr.16: 19504834-19504834 on Build GRCh38
rs572055385 Chr.16: 19504745-19504745 on Build GRCh38
rs756684532 Chr.16: 19504756-19504756 on Build GRCh38
rs756658093 Chr.16: 19504890-19504890 on Build GRCh38
rs758784142 Chr.16: 19504877-19504877 on Build GRCh38
rs879409705 Chr.16: 19504766-19504766 on Build GRCh38
rs777977948 Chr.16: 19504831-19504832 on Build GRCh38
rs780723543 Chr.16: 19504880-19504880 on Build GRCh38
rs1023458711 Chr.16: 19504885-19504885 on Build GRCh38
rs754531926 Chr.16: 19504858-19504858 on Build GRCh38
rs1005421126 Chr.16: 19504884-19504884 on Build GRCh38
rs201968694 Chr.16: 19504936-19504936 on Build GRCh38
rs779324598 Chr.16: 19504934-19504934 on Build GRCh38
rs1041083874 Chr.16: 19504804-19504804 on Build GRCh38
rs929958655 Chr.16: 19504828-19504828 on Build GRCh38
rs369924505 Chr.16: 19504928-19504928 on Build GRCh38
rs750233613 Chr.16: 19504843-19504843 on Build GRCh38
rs200327001 Chr.16: 19504881-19504881 on Build GRCh38
rs765589950 Chr.16: 19504822-19504822 on Build GRCh38
rs765815603 Chr.16: 19504844-19504847 on Build GRCh38
rs942776070 Chr.16: 19504791-19504791 on Build GRCh38
rs369215008 Chr.16: 19504740-19504740 on Build GRCh38

View Full Product Details