Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222599859-222600130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSB
Entrez Gene ID
10056
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Gene Aliases
FARSLB, FRSB, HSPC173, PheHB, PheRS
UniGene
Hs.471452
Gene Chromosome Location
Chr.2: 222571443-222656355 on Build GRCh38
Transcripts
NM_005687.4 NR_130154.1 XM_006712169.2 XM_011510466.2 XM_017003110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631862 c.1574C>T p.P525L Chr.2: 222599972-222599972 on Build GRCh38
5498826 c.1595delG p.G532fs*3 Chr.2: 222599951-222599951 on Build GRCh38
6467005 c.1534C>T p.H512Y Chr.2: 222600012-222600012 on Build GRCh38
442388 c.1595G>A p.G532E Chr.2: 222599951-222599951 on Build GRCh38
3578210 c.1598A>T p.Y533F Chr.2: 222599948-222599948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760043036 Chr.2: 222599927-222599927 on Build GRCh38
rs773284715 Chr.2: 222600038-222600038 on Build GRCh38
rs186471122 Chr.2: 222599972-222599972 on Build GRCh38
rs751321614 Chr.2: 222599984-222599984 on Build GRCh38
rs759860312 Chr.2: 222600088-222600088 on Build GRCh38
rs747176811 Chr.2: 222600060-222600060 on Build GRCh38
rs373227385 Chr.2: 222599894-222599894 on Build GRCh38
rs781076183 Chr.2: 222600001-222600001 on Build GRCh38
rs772250086 Chr.2: 222599898-222599898 on Build GRCh38
rs774439903 Chr.2: 222599888-222599888 on Build GRCh38
rs772209314 Chr.2: 222600028-222600028 on Build GRCh38
rs748247701 Chr.2: 222600026-222600026 on Build GRCh38
rs752762676 Chr.2: 222599985-222599985 on Build GRCh38
rs1054521434 Chr.2: 222599970-222599970 on Build GRCh38
rs771168861 Chr.2: 222600065-222600065 on Build GRCh38
rs866394238 Chr.2: 222599936-222599937 on Build GRCh38
rs747779783 Chr.2: 222600067-222600067 on Build GRCh38
rs752222330 Chr.2: 222599950-222599950 on Build GRCh38
rs756280936 Chr.2: 222600019-222600019 on Build GRCh38
rs936948545 Chr.2: 222599954-222599954 on Build GRCh38
rs375635278 Chr.2: 222600089-222600089 on Build GRCh38
rs756971790 Chr.2: 222599997-222599997 on Build GRCh38
rs918076712 Chr.2: 222599902-222599902 on Build GRCh38
rs780610995 Chr.2: 222600058-222600059 on Build GRCh38
rs777551426 Chr.2: 222599979-222599979 on Build GRCh38
rs369727146 Chr.2: 222599911-222599911 on Build GRCh38
rs777018523 Chr.2: 222600076-222600076 on Build GRCh38
rs754503104 Chr.2: 222599906-222599906 on Build GRCh38
rs943105963 Chr.2: 222600102-222600102 on Build GRCh38
rs765697968 Chr.2: 222599936-222599936 on Build GRCh38
rs745671879 Chr.2: 222600017-222600017 on Build GRCh38
rs1016325037 Chr.2: 222599951-222599951 on Build GRCh38
rs773483382 Chr.2: 222599903-222599903 on Build GRCh38
rs140241710 Chr.2: 222599919-222599919 on Build GRCh38
rs766664351 Chr.2: 222599912-222599912 on Build GRCh38
rs990927135 Chr.2: 222599891-222599891 on Build GRCh38

View Full Product Details