Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111762536-111762789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX20
Entrez Gene ID
11218
Gene Name
DEAD-box helicase 20
Gene Aliases
DP103, GEMIN3
UniGene
Hs.591405
Gene Chromosome Location
Chr.1: 111755568-111767577 on Build GRCh38
Transcripts
NM_007204.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895154 c.1210+2_1210+3insA p.? Chr.1: 111762784-111762785 on Build GRCh38
4020653 c.1168T>C p.W390R Chr.1: 111762740-111762740 on Build GRCh38
7270338 c.1203C>T p.G401G Chr.1: 111762775-111762775 on Build GRCh38
3376645 c.1162T>C p.L388L Chr.1: 111762734-111762734 on Build GRCh38
1735966 c.1210+3_1210+4delAA p.? Chr.1: 111762785-111762786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527851045 Chr.1: 111762618-111762618 on Build GRCh38
rs519155 Chr.1: 111762606-111762606 on Build GRCh38
rs61755354 Chr.1: 111762683-111762683 on Build GRCh38
rs903060596 Chr.1: 111762610-111762610 on Build GRCh38
rs757896588 Chr.1: 111762673-111762673 on Build GRCh38
rs748215926 Chr.1: 111762747-111762747 on Build GRCh38
rs971869269 Chr.1: 111762752-111762752 on Build GRCh38
rs961887852 Chr.1: 111762710-111762710 on Build GRCh38
rs770140983 Chr.1: 111762629-111762629 on Build GRCh38
rs752201060 Chr.1: 111762672-111762672 on Build GRCh38
rs185843735 Chr.1: 111762620-111762620 on Build GRCh38
rs562319850 Chr.1: 111762607-111762607 on Build GRCh38
rs771207510 Chr.1: 111762762-111762762 on Build GRCh38
rs772230467 Chr.1: 111762656-111762656 on Build GRCh38
rs769139383 Chr.1: 111762641-111762641 on Build GRCh38
rs191486012 Chr.1: 111762622-111762622 on Build GRCh38
rs761411621 Chr.1: 111762713-111762715 on Build GRCh38
rs532804198 Chr.1: 111762723-111762723 on Build GRCh38
rs760668879 Chr.1: 111762631-111762631 on Build GRCh38
rs373863647 Chr.1: 111762569-111762569 on Build GRCh38
rs759581160 Chr.1: 111762670-111762670 on Build GRCh38
rs773622704 Chr.1: 111762756-111762756 on Build GRCh38
rs915695352 Chr.1: 111762561-111762561 on Build GRCh38
rs377458578 Chr.1: 111762727-111762727 on Build GRCh38
rs760611647 Chr.1: 111762660-111762660 on Build GRCh38
rs748208565 Chr.1: 111762651-111762651 on Build GRCh38
rs371543712 Chr.1: 111762650-111762650 on Build GRCh38
rs137956268 Chr.1: 111762601-111762601 on Build GRCh38
rs776506696 Chr.1: 111762668-111762668 on Build GRCh38
rs1052720949 Chr.1: 111762623-111762623 on Build GRCh38
rs376030290 Chr.1: 111762632-111762632 on Build GRCh38
rs779452871 Chr.1: 111762730-111762730 on Build GRCh38
rs373513671 Chr.1: 111762671-111762671 on Build GRCh38
rs755461226 Chr.1: 111762726-111762726 on Build GRCh38
rs753097408 Chr.1: 111762666-111762670 on Build GRCh38
rs766258594 Chr.1: 111762665-111762665 on Build GRCh38
rs373018178 Chr.1: 111762649-111762649 on Build GRCh38
rs763722623 Chr.1: 111762684-111762684 on Build GRCh38
rs996532842 Chr.1: 111762648-111762648 on Build GRCh38
rs567458107 Chr.1: 111762565-111762565 on Build GRCh38
rs562883339 Chr.1: 111762595-111762595 on Build GRCh38
rs772184099 Chr.1: 111762749-111762749 on Build GRCh38
rs773533148 Chr.1: 111762659-111762659 on Build GRCh38
rs749810208 Chr.1: 111762636-111762636 on Build GRCh38
rs143443770 Chr.1: 111762703-111762703 on Build GRCh38
rs1026970002 Chr.1: 111762725-111762725 on Build GRCh38
rs200507788 Chr.1: 111762758-111762758 on Build GRCh38

View Full Product Details